Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100067856-100068115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD14A
Entrez Gene ID
64645
Gene Name
major facilitator superfamily domain containing 14A
Gene Aliases
HIAT1
UniGene
Hs.124156
Gene Chromosome Location
Chr.1: 100038078-100083377 on Build GRCh38
Transcripts
NM_033055.2 XM_005271118.1 XM_011541957.1 XM_017002084.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6057330 c.557G>A p.G186E Chr.1: 100068047-100068047 on Build GRCh38
4019785 c.559G>A p.D187N Chr.1: 100068049-100068049 on Build GRCh38
1209653 c.533G>A p.G178E Chr.1: 100068023-100068023 on Build GRCh38
7077073 c.529A>G p.I177V Chr.1: 100068019-100068019 on Build GRCh38
5443464 c.608G>T p.C203F Chr.1: 100068098-100068098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019973628 Chr.1: 100068051-100068051 on Build GRCh38
rs753726145 Chr.1: 100067945-100067945 on Build GRCh38
rs781342608 Chr.1: 100068020-100068020 on Build GRCh38
rs149078184 Chr.1: 100067932-100067932 on Build GRCh38
rs757729973 Chr.1: 100067952-100067955 on Build GRCh38
rs201979443 Chr.1: 100068077-100068077 on Build GRCh38
rs749701775 Chr.1: 100068079-100068079 on Build GRCh38
rs145252879 Chr.1: 100067918-100067918 on Build GRCh38
rs57251883 Chr.1: 100067971-100067971 on Build GRCh38
rs780362354 Chr.1: 100068045-100068045 on Build GRCh38
rs764177406 Chr.1: 100067958-100067958 on Build GRCh38
rs375261895 Chr.1: 100068023-100068023 on Build GRCh38
rs746359285 Chr.1: 100068037-100068037 on Build GRCh38
rs774052132 Chr.1: 100067920-100067920 on Build GRCh38
rs954612005 Chr.1: 100067949-100067949 on Build GRCh38
rs185181479 Chr.1: 100067951-100067951 on Build GRCh38
rs964395884 Chr.1: 100067908-100067908 on Build GRCh38
rs754066940 Chr.1: 100067950-100067951 on Build GRCh38
rs550795939 Chr.1: 100068062-100068062 on Build GRCh38
rs757220012 Chr.1: 100068015-100068015 on Build GRCh38
rs1017979802 Chr.1: 100067896-100067896 on Build GRCh38
rs778159377 Chr.1: 100067931-100067931 on Build GRCh38
rs779306754 Chr.1: 100067985-100067985 on Build GRCh38
rs368246120 Chr.1: 100067955-100067955 on Build GRCh38
rs200684961 Chr.1: 100068039-100068039 on Build GRCh38

View Full Product Details