Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70029375-70029648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDA
Entrez Gene ID
1896
Gene Name
ectodysplasin A
Gene Aliases
ECTD1, ED1, ED1-A1, ED1-A2, EDA-A1, EDA-A2, EDA1, EDA2, HED, HED1, ODT1, STHAGX1, TNLG7C, XHED, XLHED
UniGene
Hs.105407
Gene Chromosome Location
Chr.X: 69616067-70039472 on Build GRCh38
Transcripts
NM_001005609.1 NM_001005612.2 NM_001399.4 XM_006724630.2 XM_011530885.2 XM_017029336.1

SNP Detail

SNP ID Chromosome Location
rs182401776 Chr.X: 70029411-70029411 on Build GRCh38
rs369843699 Chr.X: 70029495-70029495 on Build GRCh38
rs372225521 Chr.X: 70029439-70029439 on Build GRCh38
rs199852492 Chr.X: 70029556-70029556 on Build GRCh38
rs765962085 Chr.X: 70029565-70029565 on Build GRCh38
rs768962573 Chr.X: 70029476-70029476 on Build GRCh38
rs751258727 Chr.X: 70029547-70029547 on Build GRCh38
rs886039466 Chr.X: 70029503-70029503 on Build GRCh38
rs772528585 Chr.X: 70029489-70029489 on Build GRCh38
rs889471252 Chr.X: 70029481-70029481 on Build GRCh38
rs761304256 Chr.X: 70029499-70029499 on Build GRCh38
rs762249660 Chr.X: 70029486-70029486 on Build GRCh38
rs375446877 Chr.X: 70029470-70029470 on Build GRCh38
rs397516672 Chr.X: 70029527-70029527 on Build GRCh38
rs756966486 Chr.X: 70029555-70029555 on Build GRCh38
rs763687144 Chr.X: 70029543-70029543 on Build GRCh38
rs202225741 Chr.X: 70029587-70029587 on Build GRCh38
rs762479527 Chr.X: 70029542-70029542 on Build GRCh38
rs752134804 Chr.X: 70029535-70029535 on Build GRCh38
rs774740976 Chr.X: 70029485-70029485 on Build GRCh38
rs140058036 Chr.X: 70029585-70029585 on Build GRCh38
rs756015428 Chr.X: 70029564-70029564 on Build GRCh38
rs766810445 Chr.X: 70029514-70029514 on Build GRCh38
rs397516673 Chr.X: 70029537-70029537 on Build GRCh38
rs886042183 Chr.X: 70029538-70029538 on Build GRCh38

View Full Product Details