Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 94127656-94127907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFI27L2
Entrez Gene ID
83982
Gene Name
interferon alpha inducible protein 27 like 2
Gene Aliases
FAM14A, ISG12B, TLH29
UniGene
Hs.94695
Gene Chromosome Location
Chr.14: 94127781-94129620 on Build GRCh38
Transcripts
NM_032036.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5439866 c.385G>A p.E129K Chr.14: 94127807-94127807 on Build GRCh38
959123 c.304C>A p.P102T Chr.14: 94127888-94127888 on Build GRCh38
5016459 c.322delG p.E108fs*>23 Chr.14: 94127870-94127870 on Build GRCh38
959122 c.388G>A p.E130K Chr.14: 94127804-94127804 on Build GRCh38
5012821 c.321T>A p.D107E Chr.14: 94127871-94127871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763184903 Chr.14: 94127863-94127863 on Build GRCh38
rs765926288 Chr.14: 94127798-94127798 on Build GRCh38
rs774303942 Chr.14: 94127877-94127877 on Build GRCh38
rs771753231 Chr.14: 94127751-94127751 on Build GRCh38
rs772737094 Chr.14: 94127871-94127871 on Build GRCh38
rs368024326 Chr.14: 94127760-94127760 on Build GRCh38
rs780325177 Chr.14: 94127809-94127809 on Build GRCh38
rs1052113215 Chr.14: 94127754-94127754 on Build GRCh38
rs140775821 Chr.14: 94127726-94127726 on Build GRCh38
rs911083 Chr.14: 94127818-94127818 on Build GRCh38
rs746286381 Chr.14: 94127827-94127827 on Build GRCh38
rs927774766 Chr.14: 94127699-94127699 on Build GRCh38
rs144628066 Chr.14: 94127802-94127802 on Build GRCh38
rs551090604 Chr.14: 94127761-94127761 on Build GRCh38
rs771345141 Chr.14: 94127796-94127796 on Build GRCh38
rs143459295 Chr.14: 94127845-94127845 on Build GRCh38
rs528632867 Chr.14: 94127693-94127693 on Build GRCh38
rs766282299 Chr.14: 94127886-94127886 on Build GRCh38
rs764122808 Chr.14: 94127876-94127876 on Build GRCh38
rs146463622 Chr.14: 94127697-94127697 on Build GRCh38
rs571551864 Chr.14: 94127706-94127706 on Build GRCh38
rs113306298 Chr.14: 94127885-94127885 on Build GRCh38
rs1022398364 Chr.14: 94127786-94127786 on Build GRCh38
rs144242035 Chr.14: 94127860-94127860 on Build GRCh38
rs751140047 Chr.14: 94127872-94127874 on Build GRCh38
rs1011393321 Chr.14: 94127767-94127767 on Build GRCh38
rs555343689 Chr.14: 94127852-94127852 on Build GRCh38
rs200222730 Chr.14: 94127775-94127775 on Build GRCh38
rs938975975 Chr.14: 94127745-94127745 on Build GRCh38

View Full Product Details