Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49815714-49815987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAT7
Entrez Gene ID
11143
Gene Name
lysine acetyltransferase 7
Gene Aliases
HBO1, HBOA, MYST2, ZC2HC7
UniGene
Hs.21907
Gene Chromosome Location
Chr.17: 49788619-49829096 on Build GRCh38
Transcripts
NM_001199155.1 NM_001199156.1 NM_001199157.1 NM_001199158.1 NM_007067.4 XM_011524235.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4067501 c.925C>A p.L309I Chr.17: 49815875-49815875 on Build GRCh38
75541 c.935G>C p.R312T Chr.17: 49815885-49815885 on Build GRCh38
2698476 c.930C>T p.F310F Chr.17: 49815880-49815880 on Build GRCh38
1302983 c.910G>A p.E304K Chr.17: 49815860-49815860 on Build GRCh38
3518948 c.867C>T p.T289T Chr.17: 49815817-49815817 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372076989 Chr.17: 49815816-49815816 on Build GRCh38
rs767673655 Chr.17: 49815933-49815933 on Build GRCh38
rs778321388 Chr.17: 49815777-49815777 on Build GRCh38
rs748512120 Chr.17: 49815755-49815755 on Build GRCh38
rs773298063 Chr.17: 49815859-49815859 on Build GRCh38
rs761764906 Chr.17: 49815839-49815839 on Build GRCh38
rs201695121 Chr.17: 49815897-49815897 on Build GRCh38
rs373608261 Chr.17: 49815961-49815961 on Build GRCh38
rs980211837 Chr.17: 49815872-49815872 on Build GRCh38
rs763062488 Chr.17: 49815868-49815868 on Build GRCh38
rs779593715 Chr.17: 49815951-49815951 on Build GRCh38
rs1005858344 Chr.17: 49815745-49815745 on Build GRCh38
rs756583008 Chr.17: 49815763-49815763 on Build GRCh38
rs938711194 Chr.17: 49815921-49815921 on Build GRCh38
rs983244954 Chr.17: 49815773-49815773 on Build GRCh38
rs746537771 Chr.17: 49815958-49815958 on Build GRCh38
rs768346284 Chr.17: 49815960-49815960 on Build GRCh38
rs565097216 Chr.17: 49815944-49815944 on Build GRCh38
rs776452039 Chr.17: 49815830-49815830 on Build GRCh38
rs771573782 Chr.17: 49815783-49815783 on Build GRCh38
rs757854135 Chr.17: 49815948-49815948 on Build GRCh38
rs909083351 Chr.17: 49815779-49815779 on Build GRCh38
rs746696166 Chr.17: 49815793-49815793 on Build GRCh38
rs1057184846 Chr.17: 49815927-49815927 on Build GRCh38
rs753103174 Chr.17: 49815941-49815941 on Build GRCh38
rs139974135 Chr.17: 49815856-49815856 on Build GRCh38
rs755206820 Chr.17: 49815932-49815932 on Build GRCh38
rs927260683 Chr.17: 49815904-49815904 on Build GRCh38
rs1035477939 Chr.17: 49815869-49815869 on Build GRCh38
rs368177767 Chr.17: 49815784-49815784 on Build GRCh38
rs751687487 Chr.17: 49815906-49815906 on Build GRCh38
rs745371078 Chr.17: 49815778-49815778 on Build GRCh38
rs117174322 Chr.17: 49815943-49815943 on Build GRCh38
rs950658424 Chr.17: 49815771-49815771 on Build GRCh38

View Full Product Details