Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 124952221-124952446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX37
Entrez Gene ID
57647
Gene Name
DEAH-box helicase 37
Gene Aliases
DDX37
UniGene
Hs.107382
Gene Chromosome Location
Chr.12: 124946824-124989125 on Build GRCh38
Transcripts
NM_032656.3 XM_005253590.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1203553 c.2820G>T p.R940S Chr.12: 124952446-124952446 on Build GRCh38
4039977 c.2859C>T p.N953N Chr.12: 124952407-124952407 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs373457030 Chr.12: 124952372-124952372 on Build GRCh38
rs552353272 Chr.12: 124952418-124952418 on Build GRCh38
rs770324345 Chr.12: 124952395-124952395 on Build GRCh38
rs767211430 Chr.12: 124952427-124952427 on Build GRCh38
rs777335012 Chr.12: 124952384-124952384 on Build GRCh38
rs1021729863 Chr.12: 124952341-124952341 on Build GRCh38
rs4502074 Chr.12: 124952351-124952351 on Build GRCh38
rs80318053 Chr.12: 124952356-124952356 on Build GRCh38
rs775956694 Chr.12: 124952407-124952407 on Build GRCh38
rs748356878 Chr.12: 124952359-124952359 on Build GRCh38
rs370278054 Chr.12: 124952421-124952421 on Build GRCh38
rs765281640 Chr.12: 124952371-124952371 on Build GRCh38
rs200048716 Chr.12: 124952393-124952393 on Build GRCh38
rs937168238 Chr.12: 124952273-124952273 on Build GRCh38
rs561787414 Chr.12: 124952323-124952323 on Build GRCh38
rs747120066 Chr.12: 124952381-124952381 on Build GRCh38
rs377438294 Chr.12: 124952428-124952428 on Build GRCh38
rs777660625 Chr.12: 124952380-124952380 on Build GRCh38
rs1004971672 Chr.12: 124952376-124952376 on Build GRCh38
rs564291798 Chr.12: 124952268-124952268 on Build GRCh38
rs753621820 Chr.12: 124952350-124952350 on Build GRCh38
rs531372766 Chr.12: 124952284-124952284 on Build GRCh38
rs963571003 Chr.12: 124952391-124952391 on Build GRCh38
rs545620991 Chr.12: 124952252-124952252 on Build GRCh38
rs4375522 Chr.12: 124952304-124952304 on Build GRCh38
rs771564965 Chr.12: 124952383-124952383 on Build GRCh38
rs759957909 Chr.12: 124952394-124952394 on Build GRCh38

View Full Product Details