Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 196914540-196914805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFHR4
Entrez Gene ID
10877
Gene Name
complement factor H related 4
Gene Aliases
CFHL4, FHR-4, FHR4
UniGene
Hs.710100
Gene Chromosome Location
Chr.1: 196888014-196918911 on Build GRCh38
Transcripts
NM_001201550.2 NM_001201551.1 NM_006684.4 XM_006711129.3 XM_017000110.1 XM_017000111.1 XM_017000112.1 XM_017000113.1 XM_017000114.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5482523 c.596C>A p.S199Y Chr.1: 196914651-196914651 on Build GRCh38
6363888 c.1229T>A p.M410K Chr.1: 196914546-196914546 on Build GRCh38
678402 c.511A>T p.T171S Chr.1: 196914566-196914566 on Build GRCh38
901585 c.536G>T p.G179V Chr.1: 196914591-196914591 on Build GRCh38
3480889 c.1254G>A p.L418L Chr.1: 196914571-196914571 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150996656 Chr.1: 196914659-196914659 on Build GRCh38
rs372504829 Chr.1: 196914651-196914651 on Build GRCh38
rs774791209 Chr.1: 196914618-196914618 on Build GRCh38
rs763585687 Chr.1: 196914615-196914615 on Build GRCh38
rs944094588 Chr.1: 196914745-196914746 on Build GRCh38
rs769399907 Chr.1: 196914603-196914603 on Build GRCh38
rs781319482 Chr.1: 196914645-196914645 on Build GRCh38
rs540676982 Chr.1: 196914697-196914697 on Build GRCh38
rs376665896 Chr.1: 196914634-196914634 on Build GRCh38
rs769806412 Chr.1: 196914646-196914646 on Build GRCh38
rs770918628 Chr.1: 196914590-196914590 on Build GRCh38
rs762274604 Chr.1: 196914654-196914654 on Build GRCh38
rs772297890 Chr.1: 196914666-196914666 on Build GRCh38
rs377192921 Chr.1: 196914677-196914677 on Build GRCh38
rs762070970 Chr.1: 196914621-196914621 on Build GRCh38
rs766807782 Chr.1: 196914684-196914684 on Build GRCh38
rs185136769 Chr.1: 196914750-196914750 on Build GRCh38
rs575526842 Chr.1: 196914600-196914600 on Build GRCh38
rs762673293 Chr.1: 196914671-196914674 on Build GRCh38
rs768413514 Chr.1: 196914650-196914650 on Build GRCh38
rs550990938 Chr.1: 196914763-196914763 on Build GRCh38
rs756233429 Chr.1: 196914629-196914629 on Build GRCh38
rs375415067 Chr.1: 196914578-196914578 on Build GRCh38
rs779714406 Chr.1: 196914647-196914647 on Build GRCh38
rs750558068 Chr.1: 196914562-196914562 on Build GRCh38
rs376896638 Chr.1: 196914648-196914648 on Build GRCh38
rs148714898 Chr.1: 196914728-196914728 on Build GRCh38
rs975897815 Chr.1: 196914747-196914747 on Build GRCh38
rs1027167947 Chr.1: 196914667-196914667 on Build GRCh38
rs762725142 Chr.1: 196914610-196914610 on Build GRCh38
rs750894613 Chr.1: 196914627-196914627 on Build GRCh38
rs756178556 Chr.1: 196914568-196914568 on Build GRCh38
rs773514113 Chr.1: 196914676-196914676 on Build GRCh38
rs974289309 Chr.1: 196914658-196914658 on Build GRCh38
rs752831043 Chr.1: 196914723-196914723 on Build GRCh38
rs892412957 Chr.1: 196914577-196914577 on Build GRCh38
rs773870559 Chr.1: 196914652-196914652 on Build GRCh38
rs746884066 Chr.1: 196914644-196914644 on Build GRCh38
rs753878319 Chr.1: 196914636-196914636 on Build GRCh38
rs368884512 Chr.1: 196914642-196914642 on Build GRCh38
rs374426095 Chr.1: 196914587-196914587 on Build GRCh38
rs982714778 Chr.1: 196914720-196914722 on Build GRCh38
rs370096580 Chr.1: 196914583-196914583 on Build GRCh38
rs766466004 Chr.1: 196914630-196914630 on Build GRCh38
rs765910252 Chr.1: 196914692-196914692 on Build GRCh38
rs759524899 Chr.1: 196914706-196914706 on Build GRCh38
rs778949394 Chr.1: 196914643-196914643 on Build GRCh38
rs1009214834 Chr.1: 196914582-196914582 on Build GRCh38
rs768133590 Chr.1: 196914626-196914626 on Build GRCh38
rs777081259 Chr.1: 196914667-196914667 on Build GRCh38
rs181092418 Chr.1: 196914607-196914607 on Build GRCh38
rs921387480 Chr.1: 196914768-196914768 on Build GRCh38
rs765563732 Chr.1: 196914718-196914718 on Build GRCh38
rs542124899 Chr.1: 196914586-196914586 on Build GRCh38
rs751104128 Chr.1: 196914717-196914718 on Build GRCh38
rs267598274 Chr.1: 196914620-196914620 on Build GRCh38
rs776420586 Chr.1: 196914595-196914595 on Build GRCh38

View Full Product Details