Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68988990-68989249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF1BP
Entrez Gene ID
26128
Gene Name
KIF1 binding protein
Gene Aliases
KBP, KIAA1279, TTC20
UniGene
Hs.279580
Gene Chromosome Location
Chr.10: 68988721-69016983 on Build GRCh38
Transcripts
NM_015634.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4891643 c.349C>A p.H117N Chr.10: 68989181-68989181 on Build GRCh38
6567736 c.392C>T p.S131L Chr.10: 68989224-68989224 on Build GRCh38
6583055 c.163G>C p.E55Q Chr.10: 68988995-68988995 on Build GRCh38
3751905 c.196G>A p.G66S Chr.10: 68989028-68989028 on Build GRCh38
5448035 c.176A>G p.E59G Chr.10: 68989008-68989008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776484242 Chr.10: 68989156-68989156 on Build GRCh38
rs774462618 Chr.10: 68989020-68989020 on Build GRCh38
rs372695759 Chr.10: 68989199-68989199 on Build GRCh38
rs192748736 Chr.10: 68989084-68989084 on Build GRCh38
rs371300766 Chr.10: 68989014-68989014 on Build GRCh38
rs992870301 Chr.10: 68989101-68989101 on Build GRCh38
rs138264661 Chr.10: 68989162-68989162 on Build GRCh38
rs751092379 Chr.10: 68989211-68989211 on Build GRCh38
rs769531440 Chr.10: 68989179-68989179 on Build GRCh38
rs765711575 Chr.10: 68989156-68989173 on Build GRCh38
rs121434515 Chr.10: 68989082-68989082 on Build GRCh38
rs912912274 Chr.10: 68989149-68989149 on Build GRCh38
rs751053061 Chr.10: 68989051-68989051 on Build GRCh38
rs753204138 Chr.10: 68989217-68989217 on Build GRCh38
rs760565471 Chr.10: 68989027-68989027 on Build GRCh38
rs771149799 Chr.10: 68989090-68989090 on Build GRCh38
rs759061022 Chr.10: 68989212-68989212 on Build GRCh38
rs759829845 Chr.10: 68989069-68989071 on Build GRCh38
rs2255607 Chr.10: 68989028-68989028 on Build GRCh38
rs577504870 Chr.10: 68989174-68989174 on Build GRCh38
rs756503910 Chr.10: 68989066-68989066 on Build GRCh38
rs756665117 Chr.10: 68989219-68989219 on Build GRCh38
rs994944778 Chr.10: 68989009-68989009 on Build GRCh38
rs1030239043 Chr.10: 68989183-68989183 on Build GRCh38
rs1012864536 Chr.10: 68989046-68989046 on Build GRCh38
rs121434514 Chr.10: 68989100-68989100 on Build GRCh38
rs745948039 Chr.10: 68989151-68989151 on Build GRCh38
rs768145533 Chr.10: 68989215-68989215 on Build GRCh38
rs761628784 Chr.10: 68989159-68989159 on Build GRCh38
rs149625214 Chr.10: 68989216-68989216 on Build GRCh38
rs773038576 Chr.10: 68989182-68989182 on Build GRCh38
rs201068859 Chr.10: 68989034-68989034 on Build GRCh38
rs935579771 Chr.10: 68989022-68989022 on Build GRCh38
rs768592975 Chr.10: 68989154-68989154 on Build GRCh38
rs749517204 Chr.10: 68989088-68989088 on Build GRCh38
rs953307766 Chr.10: 68989186-68989186 on Build GRCh38
rs1014883362 Chr.10: 68989094-68989094 on Build GRCh38
rs749414163 Chr.10: 68989010-68989010 on Build GRCh38
rs764872990 Chr.10: 68989031-68989031 on Build GRCh38
rs750004526 Chr.10: 68989032-68989032 on Build GRCh38
rs773512678 Chr.10: 68989007-68989007 on Build GRCh38
rs778077029 Chr.10: 68989085-68989085 on Build GRCh38
rs755612919 Chr.10: 68989062-68989062 on Build GRCh38
rs11554780 Chr.10: 68989054-68989054 on Build GRCh38
rs754180146 Chr.10: 68989043-68989044 on Build GRCh38
rs781761858 Chr.10: 68989063-68989063 on Build GRCh38
rs986393623 Chr.10: 68989203-68989203 on Build GRCh38
rs540172119 Chr.10: 68989228-68989228 on Build GRCh38
rs774361181 Chr.10: 68989127-68989127 on Build GRCh38

View Full Product Details