Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 56624298-56624559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM208A
Entrez Gene ID
23272
Gene Name
family with sequence similarity 208 member A
Gene Aliases
C3orf63, RAP140, se89-1
UniGene
Hs.116877
Gene Chromosome Location
Chr.3: 56620132-56683242 on Build GRCh38
Transcripts
NM_001112736.1 NM_015224.3 XM_005264999.1 XM_006713077.2 XM_006713078.2 XM_011533552.2 XM_011533553.2 XM_017006030.1 XM_017006031.1 XM_017006032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5465936 c.4266G>A p.Q1422Q Chr.3: 56624513-56624513 on Build GRCh38
5796080 c.3172+6T>A p.? Chr.3: 56624473-56624473 on Build GRCh38
5465935 c.3138G>A p.Q1046Q Chr.3: 56624513-56624513 on Build GRCh38
5796081 c.4300+6T>A p.? Chr.3: 56624473-56624473 on Build GRCh38
1047508 c.4293G>A p.S1431S Chr.3: 56624486-56624486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs909681176 Chr.3: 56624418-56624418 on Build GRCh38
rs559978105 Chr.3: 56624401-56624401 on Build GRCh38
rs765959642 Chr.3: 56624434-56624435 on Build GRCh38
rs776181255 Chr.3: 56624380-56624380 on Build GRCh38
rs769222976 Chr.3: 56624446-56624446 on Build GRCh38
rs780315978 Chr.3: 56624441-56624441 on Build GRCh38
rs945424563 Chr.3: 56624493-56624493 on Build GRCh38
rs774146404 Chr.3: 56624439-56624440 on Build GRCh38
rs1043835546 Chr.3: 56624499-56624499 on Build GRCh38
rs149212000 Chr.3: 56624487-56624487 on Build GRCh38
rs563721709 Chr.3: 56624337-56624337 on Build GRCh38
rs773544532 Chr.3: 56624466-56624466 on Build GRCh38
rs544001644 Chr.3: 56624325-56624325 on Build GRCh38
rs549135591 Chr.3: 56624431-56624431 on Build GRCh38
rs752502719 Chr.3: 56624505-56624516 on Build GRCh38
rs975265519 Chr.3: 56624433-56624433 on Build GRCh38
rs539285615 Chr.3: 56624381-56624381 on Build GRCh38
rs962518809 Chr.3: 56624425-56624425 on Build GRCh38
rs149363597 Chr.3: 56624413-56624413 on Build GRCh38
rs892541455 Chr.3: 56624333-56624333 on Build GRCh38
rs759208694 Chr.3: 56624469-56624469 on Build GRCh38
rs764829852 Chr.3: 56624483-56624483 on Build GRCh38
rs1017976693 Chr.3: 56624329-56624329 on Build GRCh38
rs903984618 Chr.3: 56624515-56624515 on Build GRCh38
rs1033935326 Chr.3: 56624372-56624372 on Build GRCh38
rs867705035 Chr.3: 56624442-56624442 on Build GRCh38
rs766008327 Chr.3: 56624335-56624335 on Build GRCh38
rs542859306 Chr.3: 56624521-56624521 on Build GRCh38
rs201657342 Chr.3: 56624489-56624489 on Build GRCh38
rs762517549 Chr.3: 56624433-56624434 on Build GRCh38
rs760152594 Chr.3: 56624334-56624334 on Build GRCh38
rs193208472 Chr.3: 56624371-56624371 on Build GRCh38
rs2291499 Chr.3: 56624453-56624453 on Build GRCh38
rs1013922429 Chr.3: 56624395-56624395 on Build GRCh38
rs757226862 Chr.3: 56624518-56624518 on Build GRCh38
rs770097257 Chr.3: 56624440-56624440 on Build GRCh38
rs202213634 Chr.3: 56624472-56624472 on Build GRCh38
rs751515469 Chr.3: 56624505-56624505 on Build GRCh38
rs772466594 Chr.3: 56624457-56624457 on Build GRCh38
rs781127177 Chr.3: 56624428-56624428 on Build GRCh38
rs951189249 Chr.3: 56624409-56624409 on Build GRCh38
rs759587986 Chr.3: 56624445-56624447 on Build GRCh38
rs984362865 Chr.3: 56624379-56624379 on Build GRCh38
rs182615098 Chr.3: 56624383-56624383 on Build GRCh38
rs750207156 Chr.3: 56624522-56624522 on Build GRCh38
rs35208241 Chr.3: 56624468-56624469 on Build GRCh38
rs150032629 Chr.3: 56624486-56624486 on Build GRCh38
rs774781087 Chr.3: 56624447-56624447 on Build GRCh38
rs557390655 Chr.3: 56624382-56624382 on Build GRCh38
rs767154706 Chr.3: 56624449-56624450 on Build GRCh38

View Full Product Details