Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161727296-161727565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRLB
Entrez Gene ID
127943
Gene Name
Fc receptor like B
Gene Aliases
FCRL2, FCRLM2, FCRLY, FREB-2, FcRY
UniGene
Hs.517422
Gene Chromosome Location
Chr.1: 161721544-161728143 on Build GRCh38
Transcripts
NM_001002901.3 NM_001288829.1 NM_001288830.1 NM_001288831.1 NM_001288832.1 NM_001320241.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4684274 c.937C>T p.P313S Chr.1: 161727318-161727318 on Build GRCh38
1335703 c.1047G>A p.A349A Chr.1: 161727428-161727428 on Build GRCh38
676616 c.1155G>T p.L385L Chr.1: 161727536-161727536 on Build GRCh38
3997032 c.1148G>A p.G383D Chr.1: 161727529-161727529 on Build GRCh38
4620924 c.929G>A p.G310D Chr.1: 161727310-161727310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903250180 Chr.1: 161727421-161727421 on Build GRCh38
rs748661891 Chr.1: 161727519-161727519 on Build GRCh38
rs145348831 Chr.1: 161727414-161727414 on Build GRCh38
rs745325491 Chr.1: 161727476-161727476 on Build GRCh38
rs762296785 Chr.1: 161727320-161727320 on Build GRCh38
rs777606302 Chr.1: 161727444-161727444 on Build GRCh38
rs756871182 Chr.1: 161727454-161727454 on Build GRCh38
rs200227833 Chr.1: 161727405-161727405 on Build GRCh38
rs922740553 Chr.1: 161727353-161727353 on Build GRCh38
rs16858529 Chr.1: 161727359-161727359 on Build GRCh38
rs1038116888 Chr.1: 161727409-161727409 on Build GRCh38
rs777472750 Chr.1: 161727382-161727382 on Build GRCh38
rs770543893 Chr.1: 161727386-161727386 on Build GRCh38
rs772135463 Chr.1: 161727498-161727500 on Build GRCh38
rs986101792 Chr.1: 161727376-161727376 on Build GRCh38
rs781141039 Chr.1: 161727378-161727378 on Build GRCh38
rs773850623 Chr.1: 161727389-161727389 on Build GRCh38
rs748969312 Chr.1: 161727383-161727383 on Build GRCh38
rs201828972 Chr.1: 161727524-161727524 on Build GRCh38
rs1001483632 Chr.1: 161727479-161727479 on Build GRCh38
rs769248738 Chr.1: 161727395-161727395 on Build GRCh38
rs1010920718 Chr.1: 161727510-161727510 on Build GRCh38
rs977177213 Chr.1: 161727434-161727434 on Build GRCh38
rs868762276 Chr.1: 161727365-161727365 on Build GRCh38
rs756106933 Chr.1: 161727436-161727436 on Build GRCh38
rs200143127 Chr.1: 161727542-161727542 on Build GRCh38
rs750635850 Chr.1: 161727349-161727349 on Build GRCh38
rs751617464 Chr.1: 161727424-161727424 on Build GRCh38
rs771687860 Chr.1: 161727489-161727489 on Build GRCh38
rs747891882 Chr.1: 161727380-161727380 on Build GRCh38
rs189914946 Chr.1: 161727412-161727412 on Build GRCh38
rs557205758 Chr.1: 161727325-161727325 on Build GRCh38
rs966749684 Chr.1: 161727322-161727322 on Build GRCh38
rs940376645 Chr.1: 161727339-161727339 on Build GRCh38
rs1042223942 Chr.1: 161727418-161727418 on Build GRCh38
rs752596771 Chr.1: 161727433-161727433 on Build GRCh38
rs925634623 Chr.1: 161727494-161727494 on Build GRCh38
rs1045555761 Chr.1: 161727450-161727450 on Build GRCh38
rs200652437 Chr.1: 161727401-161727401 on Build GRCh38
rs895182682 Chr.1: 161727499-161727499 on Build GRCh38
rs755070563 Chr.1: 161727430-161727430 on Build GRCh38
rs751604139 Chr.1: 161727372-161727372 on Build GRCh38
rs542774231 Chr.1: 161727361-161727361 on Build GRCh38
rs775182231 Chr.1: 161727496-161727496 on Build GRCh38
rs766573479 Chr.1: 161727531-161727531 on Build GRCh38
rs1013176749 Chr.1: 161727474-161727474 on Build GRCh38
rs541062073 Chr.1: 161727408-161727408 on Build GRCh38
rs745316946 Chr.1: 161727485-161727486 on Build GRCh38
rs778464981 Chr.1: 161727470-161727470 on Build GRCh38
rs745378945 Chr.1: 161727392-161727392 on Build GRCh38
rs376072876 Chr.1: 161727520-161727520 on Build GRCh38
rs754844581 Chr.1: 161727377-161727377 on Build GRCh38
rs34868416 Chr.1: 161727529-161727529 on Build GRCh38
rs749054136 Chr.1: 161727445-161727445 on Build GRCh38
rs762460255 Chr.1: 161727404-161727404 on Build GRCh38
rs758365312 Chr.1: 161727492-161727492 on Build GRCh38
rs755884042 Chr.1: 161727381-161727381 on Build GRCh38
rs929705920 Chr.1: 161727432-161727432 on Build GRCh38
rs774532819 Chr.1: 161727541-161727541 on Build GRCh38
rs767631255 Chr.1: 161727431-161727431 on Build GRCh38

View Full Product Details