Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 36438659-36438910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNA1
Entrez Gene ID
8900
Gene Name
cyclin A1
Gene Aliases
CT146
UniGene
Hs.417050
Gene Chromosome Location
Chr.13: 36430488-36442882 on Build GRCh38
Transcripts
NM_001111045.1 NM_001111046.1 NM_001111047.1 NM_003914.3 XM_011535294.2 XM_011535295.2 XM_011535296.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1607046 c.785C>T p.A262V Chr.13: 36438759-36438759 on Build GRCh38
3360079 c.715A>C p.I239L Chr.13: 36438689-36438689 on Build GRCh38
6876121 c.739C>T p.L247L Chr.13: 36438713-36438713 on Build GRCh38
4147699 c.700A>T p.K234* Chr.13: 36438674-36438674 on Build GRCh38
6876122 c.813C>T p.F271F Chr.13: 36438787-36438787 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202149829 Chr.13: 36438741-36438741 on Build GRCh38
rs757570478 Chr.13: 36438811-36438811 on Build GRCh38
rs780538544 Chr.13: 36438735-36438736 on Build GRCh38
rs758038956 Chr.13: 36438732-36438736 on Build GRCh38
rs1039612606 Chr.13: 36438702-36438702 on Build GRCh38
rs751861130 Chr.13: 36438799-36438799 on Build GRCh38
rs761509086 Chr.13: 36438752-36438752 on Build GRCh38
rs78810484 Chr.13: 36438729-36438729 on Build GRCh38
rs772994512 Chr.13: 36438756-36438756 on Build GRCh38
rs765233671 Chr.13: 36438705-36438705 on Build GRCh38
rs778299617 Chr.13: 36438868-36438868 on Build GRCh38
rs376677628 Chr.13: 36438683-36438683 on Build GRCh38
rs756642822 Chr.13: 36438854-36438854 on Build GRCh38
rs144283302 Chr.13: 36438778-36438778 on Build GRCh38
rs780662270 Chr.13: 36438861-36438861 on Build GRCh38
rs550633524 Chr.13: 36438822-36438822 on Build GRCh38
rs769253231 Chr.13: 36438703-36438703 on Build GRCh38
rs747732303 Chr.13: 36438877-36438877 on Build GRCh38
rs902897015 Chr.13: 36438835-36438835 on Build GRCh38
rs200577481 Chr.13: 36438732-36438732 on Build GRCh38
rs150705387 Chr.13: 36438865-36438865 on Build GRCh38
rs763130840 Chr.13: 36438783-36438783 on Build GRCh38
rs779393715 Chr.13: 36438736-36438736 on Build GRCh38
rs183736861 Chr.13: 36438781-36438781 on Build GRCh38
rs61755283 Chr.13: 36438694-36438694 on Build GRCh38
rs752851752 Chr.13: 36438708-36438708 on Build GRCh38
rs369651921 Chr.13: 36438751-36438751 on Build GRCh38
rs770432581 Chr.13: 36438735-36438735 on Build GRCh38
rs751836659 Chr.13: 36438719-36438719 on Build GRCh38
rs748854682 Chr.13: 36438738-36438738 on Build GRCh38
rs140782401 Chr.13: 36438726-36438727 on Build GRCh38
rs766132764 Chr.13: 36438763-36438763 on Build GRCh38
rs750852987 Chr.13: 36438850-36438850 on Build GRCh38
rs778102032 Chr.13: 36438717-36438717 on Build GRCh38
rs147834946 Chr.13: 36438838-36438838 on Build GRCh38
rs764214378 Chr.13: 36438787-36438787 on Build GRCh38
rs767412507 Chr.13: 36438691-36438691 on Build GRCh38
rs757758557 Chr.13: 36438726-36438726 on Build GRCh38
rs758497518 Chr.13: 36438709-36438709 on Build GRCh38
rs1041438719 Chr.13: 36438792-36438792 on Build GRCh38
rs750419153 Chr.13: 36438693-36438693 on Build GRCh38
rs267603811 Chr.13: 36438755-36438755 on Build GRCh38
rs774093184 Chr.13: 36438746-36438746 on Build GRCh38
rs145298804 Chr.13: 36438867-36438867 on Build GRCh38

View Full Product Details