Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 76995126-76995365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLN5
Entrez Gene ID
1203
Gene Name
ceroid-lipofuscinosis, neuronal 5
Gene Aliases
NCL
UniGene
Hs.30213
Gene Chromosome Location
Chr.13: 76991924-77002517 on Build GRCh38
Transcripts
NM_006493.2 XM_011534917.2 XM_017020538.1

Gene Symbol
FBXL3
Entrez Gene ID
26224
Gene Name
F-box and leucine rich repeat protein 3
Gene Aliases
FBL3, FBL3A, FBXL3A
UniGene
Hs.508284
Gene Chromosome Location
Chr.13: 76992597-77027196 on Build GRCh38
Transcripts
NM_006493.2 XM_011534917.2 XM_017020538.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
697377 c.400G>T p.V134F Chr.13: 76995142-76995142 on Build GRCh38
6139626 c.413A>G p.D138G Chr.13: 76995155-76995155 on Build GRCh38
1177056 c.434G>A p.R145Q Chr.13: 76995176-76995176 on Build GRCh38
7208931 c.438A>G p.L146L Chr.13: 76995180-76995180 on Build GRCh38
6630561 c.433C>T p.R145* Chr.13: 76995175-76995175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs996238256 Chr.13: 76995274-76995274 on Build GRCh38
rs978026182 Chr.13: 76995332-76995332 on Build GRCh38
rs1022330558 Chr.13: 76995289-76995289 on Build GRCh38
rs779923225 Chr.13: 76995243-76995243 on Build GRCh38
rs754911333 Chr.13: 76995253-76995253 on Build GRCh38
rs1468101 Chr.13: 76995234-76995234 on Build GRCh38
rs768282295 Chr.13: 76995150-76995150 on Build GRCh38
rs147214555 Chr.13: 76995228-76995228 on Build GRCh38
rs564473065 Chr.13: 76995319-76995319 on Build GRCh38
rs555805439 Chr.13: 76995305-76995305 on Build GRCh38
rs751611392 Chr.13: 76995222-76995222 on Build GRCh38
rs138110438 Chr.13: 76995154-76995154 on Build GRCh38
rs151120510 Chr.13: 76995308-76995308 on Build GRCh38
rs545963820 Chr.13: 76995314-76995314 on Build GRCh38
rs771803882 Chr.13: 76995166-76995166 on Build GRCh38
rs200187743 Chr.13: 76995256-76995256 on Build GRCh38
rs1048147140 Chr.13: 76995193-76995193 on Build GRCh38
rs749257423 Chr.13: 76995280-76995284 on Build GRCh38
rs747732275 Chr.13: 76995259-76995259 on Build GRCh38
rs757842432 Chr.13: 76995260-76995260 on Build GRCh38
rs753855242 Chr.13: 76995245-76995245 on Build GRCh38
rs747775956 Chr.13: 76995157-76995157 on Build GRCh38
rs143759121 Chr.13: 76995185-76995185 on Build GRCh38
rs898862493 Chr.13: 76995159-76995159 on Build GRCh38
rs965174436 Chr.13: 76995268-76995268 on Build GRCh38
rs201068201 Chr.13: 76995176-76995176 on Build GRCh38
rs1009155401 Chr.13: 76995251-76995251 on Build GRCh38
rs918518741 Chr.13: 76995313-76995314 on Build GRCh38
rs745473360 Chr.13: 76995208-76995208 on Build GRCh38
rs202146713 Chr.13: 76995233-76995233 on Build GRCh38
rs772707642 Chr.13: 76995172-76995172 on Build GRCh38
rs755999018 Chr.13: 76995239-76995239 on Build GRCh38
rs750610277 Chr.13: 76995232-76995232 on Build GRCh38
rs368672718 Chr.13: 76995212-76995212 on Build GRCh38
rs372785489 Chr.13: 76995223-76995223 on Build GRCh38
rs386833971 Chr.13: 76995175-76995175 on Build GRCh38

View Full Product Details