Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35010257-35010490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRAMD1A
Entrez Gene ID
57655
Gene Name
GRAM domain containing 1A
Gene Aliases
KIAA1533
UniGene
Hs.515351
Gene Chromosome Location
Chr.19: 34994784-35026471 on Build GRCh38
Transcripts
NM_001136199.2 NM_001320034.1 NM_001320035.1 NM_001320036.1 NM_020895.4 XM_011527149.1 XM_011527153.1 XM_011527154.2 XM_011527155.1 XM_011527156.1 XM_017027034.1 XM_017027035.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5056526 c.519C>T p.S173S Chr.19: 35010373-35010373 on Build GRCh38
5389389 c.448G>A p.E150K Chr.19: 35010302-35010302 on Build GRCh38
4549287 c.469G>A p.E157K Chr.19: 35010323-35010323 on Build GRCh38
1392777 c.512C>T p.T171M Chr.19: 35010366-35010366 on Build GRCh38
4441708 c.493A>G p.N165D Chr.19: 35010347-35010347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376247833 Chr.19: 35010384-35010384 on Build GRCh38
rs767144146 Chr.19: 35010295-35010295 on Build GRCh38
rs746090275 Chr.19: 35010357-35010357 on Build GRCh38
rs548730268 Chr.19: 35010410-35010410 on Build GRCh38
rs749314060 Chr.19: 35010421-35010421 on Build GRCh38
rs555275792 Chr.19: 35010468-35010468 on Build GRCh38
rs760374647 Chr.19: 35010450-35010450 on Build GRCh38
rs780173196 Chr.19: 35010367-35010367 on Build GRCh38
rs765173393 Chr.19: 35010407-35010407 on Build GRCh38
rs750637701 Chr.19: 35010413-35010413 on Build GRCh38
rs570711580 Chr.19: 35010459-35010459 on Build GRCh38
rs746162956 Chr.19: 35010418-35010418 on Build GRCh38
rs372066079 Chr.19: 35010409-35010409 on Build GRCh38
rs368205726 Chr.19: 35010408-35010408 on Build GRCh38
rs755034748 Chr.19: 35010330-35010330 on Build GRCh38
rs749761264 Chr.19: 35010371-35010371 on Build GRCh38
rs778542766 Chr.19: 35010331-35010331 on Build GRCh38
rs538059805 Chr.19: 35010372-35010372 on Build GRCh38
rs765984604 Chr.19: 35010398-35010398 on Build GRCh38
rs374024352 Chr.19: 35010401-35010401 on Build GRCh38
rs769197917 Chr.19: 35010373-35010373 on Build GRCh38
rs199663006 Chr.19: 35010297-35010297 on Build GRCh38
rs773518920 Chr.19: 35010438-35010438 on Build GRCh38
rs769848875 Chr.19: 35010366-35010366 on Build GRCh38
rs755621608 Chr.19: 35010314-35010314 on Build GRCh38
rs781408055 Chr.19: 35010355-35010355 on Build GRCh38
rs1012339747 Chr.19: 35010307-35010307 on Build GRCh38
rs76289022 Chr.19: 35010349-35010349 on Build GRCh38
rs762202839 Chr.19: 35010385-35010385 on Build GRCh38
rs1045022070 Chr.19: 35010343-35010343 on Build GRCh38
rs753440845 Chr.19: 35010400-35010400 on Build GRCh38
rs766004695 Chr.19: 35010316-35010316 on Build GRCh38
rs974543130 Chr.19: 35010302-35010302 on Build GRCh38
rs772332412 Chr.19: 35010388-35010388 on Build GRCh38
rs760464639 Chr.19: 35010394-35010394 on Build GRCh38
rs748024957 Chr.19: 35010346-35010346 on Build GRCh38
rs192598874 Chr.19: 35010430-35010430 on Build GRCh38
rs114827501 Chr.19: 35010429-35010429 on Build GRCh38
rs753816402 Chr.19: 35010325-35010325 on Build GRCh38
rs547543498 Chr.19: 35010433-35010433 on Build GRCh38
rs138482777 Chr.19: 35010393-35010393 on Build GRCh38
rs756366391 Chr.19: 35010417-35010417 on Build GRCh38
rs987702215 Chr.19: 35010442-35010442 on Build GRCh38
rs771153960 Chr.19: 35010412-35010413 on Build GRCh38

View Full Product Details