Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65463578-65463819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP8
Entrez Gene ID
54878
Gene Name
dipeptidyl peptidase 8
Gene Aliases
DP8, DPRP-1, DPRP1, MST097, MSTP097, MSTP135, MSTP141
UniGene
Hs.458609
Gene Chromosome Location
Chr.15: 65442463-65517713 on Build GRCh38
Transcripts
NM_001320875.1 NM_001320876.1 NM_017743.5 NM_130434.4 NM_197960.3 NM_197961.3 NR_135485.1 NR_135486.1 XM_005254505.1 XM_011521730.1 XM_011521731.1 XM_011521732.2 XM_011521733.1 XM_011521734.1 XM_017022373.1 XM_017022374.1 XM_017022375.1 XM_017022376.1 XM_017022377.1 XM_017022378.1 XM_017022379.1 XM_017022380.1 XM_017022381.1 XM_017022382.1 XM_017022383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4533724 c.2008G>A p.G670S Chr.15: 65463772-65463772 on Build GRCh38
964065 c.1916A>T p.D639V Chr.15: 65463816-65463816 on Build GRCh38
7310036 c.2018A>T p.Q673L Chr.15: 65463762-65463762 on Build GRCh38
1727201 c.1970A>T p.Q657L Chr.15: 65463762-65463762 on Build GRCh38
4870888 c.1964A>T p.D655V Chr.15: 65463816-65463816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1040298420 Chr.15: 65463687-65463687 on Build GRCh38
rs923594204 Chr.15: 65463678-65463678 on Build GRCh38
rs3825891 Chr.15: 65463734-65463734 on Build GRCh38
rs12595213 Chr.15: 65463635-65463635 on Build GRCh38
rs905482857 Chr.15: 65463689-65463689 on Build GRCh38
rs990991496 Chr.15: 65463629-65463629 on Build GRCh38
rs780132902 Chr.15: 65463738-65463738 on Build GRCh38
rs946379941 Chr.15: 65463636-65463636 on Build GRCh38
rs747089802 Chr.15: 65463742-65463742 on Build GRCh38
rs958890393 Chr.15: 65463622-65463622 on Build GRCh38
rs61736633 Chr.15: 65463788-65463788 on Build GRCh38
rs548080372 Chr.15: 65463726-65463726 on Build GRCh38
rs569084315 Chr.15: 65463696-65463696 on Build GRCh38
rs764732665 Chr.15: 65463720-65463720 on Build GRCh38
rs369131089 Chr.15: 65463776-65463776 on Build GRCh38
rs758594957 Chr.15: 65463735-65463735 on Build GRCh38
rs776564607 Chr.15: 65463782-65463782 on Build GRCh38
rs552270623 Chr.15: 65463675-65463675 on Build GRCh38
rs1052519614 Chr.15: 65463619-65463619 on Build GRCh38
rs112569209 Chr.15: 65463728-65463728 on Build GRCh38
rs995699370 Chr.15: 65463710-65463710 on Build GRCh38
rs927503704 Chr.15: 65463603-65463603 on Build GRCh38
rs1004755294 Chr.15: 65463754-65463754 on Build GRCh38
rs779485010 Chr.15: 65463732-65463732 on Build GRCh38
rs749489221 Chr.15: 65463680-65463680 on Build GRCh38
rs887275260 Chr.15: 65463741-65463741 on Build GRCh38
rs531488500 Chr.15: 65463721-65463721 on Build GRCh38

View Full Product Details