Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51745832-51746105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FPR1
Entrez Gene ID
2357
Gene Name
formyl peptide receptor 1
Gene Aliases
FMLP, FPR
UniGene
Hs.753
Gene Chromosome Location
Chr.19: 51745770-51751897 on Build GRCh38
Transcripts
NM_001193306.1 NM_002029.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1612675 c.978G>C p.E326D Chr.19: 51746017-51746017 on Build GRCh38
5585262 c.949C>T p.P317S Chr.19: 51746046-51746046 on Build GRCh38
1612676 c.937A>G p.I313V Chr.19: 51746058-51746058 on Build GRCh38
4080698 c.952G>A p.A318T Chr.19: 51746043-51746043 on Build GRCh38
1564264 c.943G>A p.A315T Chr.19: 51746052-51746052 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758884060 Chr.19: 51745949-51745949 on Build GRCh38
rs775336229 Chr.19: 51746083-51746083 on Build GRCh38
rs570072877 Chr.19: 51746044-51746044 on Build GRCh38
rs939201330 Chr.19: 51745948-51745948 on Build GRCh38
rs386810387 Chr.19: 51745874-51745875 on Build GRCh38
rs551338523 Chr.19: 51746019-51746019 on Build GRCh38
rs771713666 Chr.19: 51745985-51745985 on Build GRCh38
rs768152901 Chr.19: 51746020-51746020 on Build GRCh38
rs1048756669 Chr.19: 51745959-51745959 on Build GRCh38
rs746295140 Chr.19: 51745891-51745891 on Build GRCh38
rs555690623 Chr.19: 51746069-51746069 on Build GRCh38
rs758693906 Chr.19: 51745894-51745894 on Build GRCh38
rs750789771 Chr.19: 51745939-51745939 on Build GRCh38
rs748019295 Chr.19: 51745899-51745899 on Build GRCh38
rs867341 Chr.19: 51745867-51745867 on Build GRCh38
rs370386175 Chr.19: 51745896-51745896 on Build GRCh38
rs773182339 Chr.19: 51745974-51745974 on Build GRCh38
rs928709664 Chr.19: 51745931-51745931 on Build GRCh38
rs776747042 Chr.19: 51745952-51745952 on Build GRCh38
rs781308895 Chr.19: 51746045-51746045 on Build GRCh38
rs749154560 Chr.19: 51746053-51746053 on Build GRCh38
rs150820622 Chr.19: 51745951-51745951 on Build GRCh38
rs780304392 Chr.19: 51745954-51745954 on Build GRCh38
rs377323607 Chr.19: 51745906-51745906 on Build GRCh38
rs762250607 Chr.19: 51746070-51746070 on Build GRCh38
rs774001245 Chr.19: 51745917-51745917 on Build GRCh38
rs769779151 Chr.19: 51745905-51745905 on Build GRCh38
rs781317931 Chr.19: 51745966-51745966 on Build GRCh38
rs775026535 Chr.19: 51745987-51745987 on Build GRCh38
rs139480916 Chr.19: 51745957-51745957 on Build GRCh38
rs375541232 Chr.19: 51746038-51746038 on Build GRCh38
rs765922024 Chr.19: 51745912-51745912 on Build GRCh38
rs546533480 Chr.19: 51745934-51745934 on Build GRCh38
rs17849971 Chr.19: 51746002-51746002 on Build GRCh38
rs756133617 Chr.19: 51746047-51746047 on Build GRCh38
rs769687769 Chr.19: 51745973-51745973 on Build GRCh38
rs749242571 Chr.19: 51745981-51745981 on Build GRCh38
rs748291297 Chr.19: 51745969-51745969 on Build GRCh38
rs757828035 Chr.19: 51745933-51745933 on Build GRCh38
rs749228839 Chr.19: 51745870-51745870 on Build GRCh38
rs750912266 Chr.19: 51746035-51746035 on Build GRCh38
rs759139987 Chr.19: 51745918-51745918 on Build GRCh38
rs199734809 Chr.19: 51745898-51745898 on Build GRCh38
rs752164694 Chr.19: 51745925-51745925 on Build GRCh38
rs201404212 Chr.19: 51746043-51746043 on Build GRCh38
rs766700447 Chr.19: 51746041-51746041 on Build GRCh38
rs778758034 Chr.19: 51746062-51746062 on Build GRCh38
rs775137735 Chr.19: 51746016-51746016 on Build GRCh38
rs867228 Chr.19: 51745958-51745958 on Build GRCh38
rs770785167 Chr.19: 51746054-51746054 on Build GRCh38
rs770689978 Chr.19: 51745919-51745920 on Build GRCh38
rs373135788 Chr.19: 51745919-51745919 on Build GRCh38
rs140083445 Chr.19: 51746052-51746052 on Build GRCh38
rs1055992277 Chr.19: 51746057-51746057 on Build GRCh38
rs974246861 Chr.19: 51745965-51745965 on Build GRCh38
rs752751969 Chr.19: 51746046-51746046 on Build GRCh38
rs759168000 Chr.19: 51745990-51745990 on Build GRCh38

View Full Product Details