Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61796234-61796461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FEN1
Entrez Gene ID
2237
Gene Name
flap structure-specific endonuclease 1
Gene Aliases
FEN-1, MF1, RAD2
UniGene
Hs.409065
Gene Chromosome Location
Chr.11: 61792637-61797244 on Build GRCh38
Transcripts
NM_004111.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
929521 c.958C>T p.R320* Chr.11: 61796319-61796319 on Build GRCh38
6420777 c.908A>G p.E303G Chr.11: 61796269-61796269 on Build GRCh38
929522 c.993C>T p.S331S Chr.11: 61796354-61796354 on Build GRCh38
3450799 c.1079C>T p.P360L Chr.11: 61796440-61796440 on Build GRCh38
6133829 c.999A>T p.Q333H Chr.11: 61796360-61796360 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753991316 Chr.11: 61796353-61796353 on Build GRCh38
rs958305586 Chr.11: 61796438-61796438 on Build GRCh38
rs756180049 Chr.11: 61796326-61796326 on Build GRCh38
rs772603170 Chr.11: 61796282-61796282 on Build GRCh38
rs181400086 Chr.11: 61796402-61796402 on Build GRCh38
rs967168730 Chr.11: 61796379-61796379 on Build GRCh38
rs776092981 Chr.11: 61796285-61796285 on Build GRCh38
rs749625547 Chr.11: 61796257-61796257 on Build GRCh38
rs1803573 Chr.11: 61796432-61796432 on Build GRCh38
rs890830240 Chr.11: 61796258-61796258 on Build GRCh38
rs752012566 Chr.11: 61796331-61796331 on Build GRCh38
rs542490356 Chr.11: 61796397-61796397 on Build GRCh38
rs769444109 Chr.11: 61796301-61796301 on Build GRCh38
rs763788064 Chr.11: 61796315-61796315 on Build GRCh38
rs780263785 Chr.11: 61796335-61796335 on Build GRCh38
rs573495657 Chr.11: 61796377-61796377 on Build GRCh38
rs1013335904 Chr.11: 61796370-61796370 on Build GRCh38
rs141650861 Chr.11: 61796323-61796323 on Build GRCh38
rs759067341 Chr.11: 61796286-61796286 on Build GRCh38
rs748648821 Chr.11: 61796277-61796277 on Build GRCh38
rs767523831 Chr.11: 61796427-61796427 on Build GRCh38
rs758718503 Chr.11: 61796376-61796376 on Build GRCh38
rs370181656 Chr.11: 61796433-61796433 on Build GRCh38
rs762754689 Chr.11: 61796313-61796313 on Build GRCh38
rs900276320 Chr.11: 61796411-61796411 on Build GRCh38
rs150508629 Chr.11: 61796403-61796403 on Build GRCh38
rs959198585 Chr.11: 61796369-61796369 on Build GRCh38
rs768399301 Chr.11: 61796407-61796407 on Build GRCh38
rs774266785 Chr.11: 61796413-61796413 on Build GRCh38
rs202130520 Chr.11: 61796325-61796325 on Build GRCh38
rs199498803 Chr.11: 61796308-61796308 on Build GRCh38
rs755294689 Chr.11: 61796355-61796355 on Build GRCh38
rs773287912 Chr.11: 61796428-61796428 on Build GRCh38
rs188983237 Chr.11: 61796259-61796259 on Build GRCh38
rs751470906 Chr.11: 61796319-61796319 on Build GRCh38
rs765400166 Chr.11: 61796324-61796324 on Build GRCh38
rs761723999 Chr.11: 61796425-61796425 on Build GRCh38
rs772515013 Chr.11: 61796367-61796367 on Build GRCh38
rs371918257 Chr.11: 61796356-61796356 on Build GRCh38

View Full Product Details