Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86464026-86464295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM3A
Entrez Gene ID
55818
Gene Name
lysine demethylase 3A
Gene Aliases
JHDM2A, JHMD2A, JMJD1, JMJD1A, TSGA
UniGene
Hs.557425
Gene Chromosome Location
Chr.2: 86440647-86492716 on Build GRCh38
Transcripts
NM_001146688.1 NM_018433.5 XM_006712051.3 XM_017004493.1 XM_017004494.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5533164 c.863C>T p.P288L Chr.2: 86464072-86464072 on Build GRCh38
6273561 c.907G>T p.G303C Chr.2: 86464116-86464116 on Build GRCh38
272842 c.993delA p.I333fs*18 Chr.2: 86464202-86464202 on Build GRCh38
3963186 c.1006G>C p.G336R Chr.2: 86464215-86464215 on Build GRCh38
2823049 c.974C>T p.S325F Chr.2: 86464183-86464183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778303812 Chr.2: 86464091-86464091 on Build GRCh38
rs774326483 Chr.2: 86464140-86464140 on Build GRCh38
rs752181163 Chr.2: 86464101-86464101 on Build GRCh38
rs763877890 Chr.2: 86464236-86464236 on Build GRCh38
rs777516495 Chr.2: 86464107-86464107 on Build GRCh38
rs966521974 Chr.2: 86464258-86464258 on Build GRCh38
rs772331838 Chr.2: 86464167-86464167 on Build GRCh38
rs34507616 Chr.2: 86464136-86464136 on Build GRCh38
rs748178530 Chr.2: 86464166-86464166 on Build GRCh38
rs12714187 Chr.2: 86464127-86464127 on Build GRCh38
rs754299435 Chr.2: 86464077-86464077 on Build GRCh38
rs752168318 Chr.2: 86464224-86464224 on Build GRCh38
rs751250448 Chr.2: 86464237-86464237 on Build GRCh38
rs141469380 Chr.2: 86464209-86464209 on Build GRCh38
rs200618359 Chr.2: 86464183-86464183 on Build GRCh38
rs553127897 Chr.2: 86464239-86464239 on Build GRCh38
rs754578416 Chr.2: 86464078-86464078 on Build GRCh38
rs369252210 Chr.2: 86464141-86464141 on Build GRCh38
rs769713981 Chr.2: 86464130-86464130 on Build GRCh38
rs757956716 Chr.2: 86464227-86464227 on Build GRCh38
rs745635517 Chr.2: 86464262-86464262 on Build GRCh38
rs746847069 Chr.2: 86464112-86464112 on Build GRCh38
rs535868641 Chr.2: 86464214-86464214 on Build GRCh38
rs770816854 Chr.2: 86464121-86464121 on Build GRCh38
rs142836097 Chr.2: 86464083-86464083 on Build GRCh38
rs146075276 Chr.2: 86464092-86464092 on Build GRCh38
rs773393617 Chr.2: 86464179-86464179 on Build GRCh38
rs761097290 Chr.2: 86464180-86464180 on Build GRCh38
rs758152638 Chr.2: 86464104-86464104 on Build GRCh38
rs777000192 Chr.2: 86464185-86464185 on Build GRCh38
rs761007296 Chr.2: 86464059-86464059 on Build GRCh38
rs952218558 Chr.2: 86464055-86464055 on Build GRCh38
rs759960433 Chr.2: 86464194-86464194 on Build GRCh38
rs567443572 Chr.2: 86464128-86464128 on Build GRCh38
rs766874823 Chr.2: 86464263-86464264 on Build GRCh38
rs765731866 Chr.2: 86464221-86464221 on Build GRCh38
rs766914007 Chr.2: 86464066-86464066 on Build GRCh38
rs756027880 Chr.2: 86464265-86464265 on Build GRCh38
rs1004844281 Chr.2: 86464143-86464143 on Build GRCh38
rs757175695 Chr.2: 86464238-86464238 on Build GRCh38
rs572283715 Chr.2: 86464270-86464270 on Build GRCh38

View Full Product Details