Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 36441041-36441314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIM1
Entrez Gene ID
51232
Gene Name
cysteine rich transmembrane BMP regulator 1
Gene Aliases
CRIM-1, S52
UniGene
Hs.699247
Gene Chromosome Location
Chr.2: 36356088-36551135 on Build GRCh38
Transcripts
NM_016441.2 XM_005264357.4 XM_011532898.2 XM_011532899.2 XM_011532901.2 XM_017004258.1 XM_017004259.1 XM_017004260.1 XM_017004261.1 XM_017004262.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5665126 c.514C>A p.P172T Chr.2: 36441266-36441266 on Build GRCh38
1020587 c.535T>C p.C179R Chr.2: 36441287-36441287 on Build GRCh38
4001662 c.506-6T>A p.? Chr.2: 36441252-36441252 on Build GRCh38
5862472 c.506-2_506-1delAG p.? Chr.2: 36441256-36441257 on Build GRCh38
6542519 c.529G>T p.A177S Chr.2: 36441281-36441281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934805289 Chr.2: 36441203-36441203 on Build GRCh38
rs1022443279 Chr.2: 36441118-36441118 on Build GRCh38
rs201280323 Chr.2: 36441282-36441282 on Build GRCh38
rs139233220 Chr.2: 36441142-36441142 on Build GRCh38
rs143852431 Chr.2: 36441154-36441154 on Build GRCh38
rs2276673 Chr.2: 36441252-36441252 on Build GRCh38
rs183690059 Chr.2: 36441153-36441153 on Build GRCh38
rs150852612 Chr.2: 36441109-36441109 on Build GRCh38
rs1049505643 Chr.2: 36441232-36441232 on Build GRCh38
rs144261626 Chr.2: 36441286-36441286 on Build GRCh38
rs762374786 Chr.2: 36441273-36441273 on Build GRCh38
rs376912980 Chr.2: 36441216-36441216 on Build GRCh38
rs2049367 Chr.2: 36441076-36441076 on Build GRCh38
rs748063745 Chr.2: 36441212-36441212 on Build GRCh38
rs138252009 Chr.2: 36441285-36441285 on Build GRCh38
rs773057254 Chr.2: 36441235-36441235 on Build GRCh38
rs932417688 Chr.2: 36441229-36441229 on Build GRCh38
rs991490485 Chr.2: 36441135-36441135 on Build GRCh38
rs1027688506 Chr.2: 36441092-36441092 on Build GRCh38
rs370583153 Chr.2: 36441250-36441250 on Build GRCh38
rs770685984 Chr.2: 36441249-36441249 on Build GRCh38
rs766992854 Chr.2: 36441267-36441267 on Build GRCh38
rs141047679 Chr.2: 36441239-36441239 on Build GRCh38
rs112482945 Chr.2: 36441194-36441194 on Build GRCh38
rs751735637 Chr.2: 36441291-36441291 on Build GRCh38
rs377616585 Chr.2: 36441276-36441276 on Build GRCh38
rs765718384 Chr.2: 36441274-36441274 on Build GRCh38
rs532895238 Chr.2: 36441073-36441073 on Build GRCh38
rs147319024 Chr.2: 36441189-36441189 on Build GRCh38
rs141493664 Chr.2: 36441101-36441101 on Build GRCh38
rs779997546 Chr.2: 36441219-36441219 on Build GRCh38
rs992916142 Chr.2: 36441211-36441211 on Build GRCh38
rs982060948 Chr.2: 36441167-36441167 on Build GRCh38
rs753705676 Chr.2: 36441067-36441067 on Build GRCh38

View Full Product Details