Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7967422-7967685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELAVL1
Entrez Gene ID
1994
Gene Name
ELAV like RNA binding protein 1
Gene Aliases
ELAV1, HUR, Hua, MelG
UniGene
Hs.184492
Gene Chromosome Location
Chr.19: 7958573-8005645 on Build GRCh38
Transcripts
NM_001419.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6653135 c.562A>G p.N188D Chr.19: 7967659-7967659 on Build GRCh38
6271889 c.636C>A p.A212A Chr.19: 7967666-7967666 on Build GRCh38
6653137 c.606G>A p.S202S Chr.19: 7967615-7967615 on Build GRCh38
2729784 c.693G>A p.A231A Chr.19: 7967609-7967609 on Build GRCh38
5013356 c.664C>T p.L222F Chr.19: 7967638-7967638 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2162798 Chr.19: 7967525-7967525 on Build GRCh38
rs370407808 Chr.19: 7967610-7967610 on Build GRCh38
rs977350335 Chr.19: 7967485-7967485 on Build GRCh38
rs201956891 Chr.19: 7967547-7967547 on Build GRCh38
rs906455857 Chr.19: 7967521-7967521 on Build GRCh38
rs571965657 Chr.19: 7967462-7967462 on Build GRCh38
rs756212952 Chr.19: 7967611-7967611 on Build GRCh38
rs967297384 Chr.19: 7967482-7967482 on Build GRCh38
rs986000514 Chr.19: 7967545-7967545 on Build GRCh38
rs112949142 Chr.19: 7967533-7967533 on Build GRCh38
rs778988164 Chr.19: 7967639-7967639 on Build GRCh38
rs906836339 Chr.19: 7967633-7967633 on Build GRCh38
rs138823708 Chr.19: 7967578-7967578 on Build GRCh38
rs377051094 Chr.19: 7967600-7967600 on Build GRCh38
rs1002541009 Chr.19: 7967454-7967454 on Build GRCh38
rs781118578 Chr.19: 7967474-7967474 on Build GRCh38
rs759812667 Chr.19: 7967546-7967546 on Build GRCh38
rs372959704 Chr.19: 7967615-7967615 on Build GRCh38
rs769836408 Chr.19: 7967520-7967520 on Build GRCh38
rs576719241 Chr.19: 7967576-7967576 on Build GRCh38
rs758108261 Chr.19: 7967591-7967591 on Build GRCh38
rs1034102127 Chr.19: 7967455-7967455 on Build GRCh38
rs143110565 Chr.19: 7967453-7967453 on Build GRCh38
rs185507558 Chr.19: 7967481-7967481 on Build GRCh38
rs919267901 Chr.19: 7967603-7967603 on Build GRCh38
rs372525627 Chr.19: 7967557-7967557 on Build GRCh38
rs151199397 Chr.19: 7967483-7967483 on Build GRCh38
rs749481743 Chr.19: 7967524-7967524 on Build GRCh38
rs774920988 Chr.19: 7967621-7967621 on Build GRCh38
rs867072121 Chr.19: 7967587-7967587 on Build GRCh38
rs3826740 Chr.19: 7967548-7967548 on Build GRCh38
rs534636857 Chr.19: 7967463-7967463 on Build GRCh38
rs746289699 Chr.19: 7967644-7967644 on Build GRCh38
rs750293457 Chr.19: 7967590-7967590 on Build GRCh38
rs370080641 Chr.19: 7967536-7967536 on Build GRCh38
rs555253806 Chr.19: 7967452-7967452 on Build GRCh38
rs942136957 Chr.19: 7967553-7967553 on Build GRCh38
rs1046523870 Chr.19: 7967450-7967450 on Build GRCh38
rs756868982 Chr.19: 7967577-7967577 on Build GRCh38
rs772532543 Chr.19: 7967645-7967645 on Build GRCh38
rs763656994 Chr.19: 7967558-7967558 on Build GRCh38
rs916382705 Chr.19: 7967479-7967479 on Build GRCh38
rs201498939 Chr.19: 7967663-7967663 on Build GRCh38

View Full Product Details