Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 105506424-105506649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSD1L
Entrez Gene ID
83856
Gene Name
fibronectin type III and SPRY domain containing 1 like
Gene Aliases
CCDC10, CSDUFD1, FSD1CL, FSD1NL, MIR1
UniGene
Hs.136901
Gene Chromosome Location
Chr.9: 105442183-105552433 on Build GRCh38
Transcripts
NM_001145313.2 NM_001287191.1 NM_001287192.1 XM_005252254.2 XM_005252257.3 XM_005252260.3 XM_011519077.2 XM_011519078.2 XM_011519079.2 XM_011519080.2 XM_011519081.2 XM_017015182.1 XM_017015183.1 XM_017015184.1 XM_017015185.1 XM_017015186.1 XM_017015187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5003939 c.260G>A p.R87H Chr.9: 105506543-105506543 on Build GRCh38
1103566 c.667G>A p.E223K Chr.9: 105506479-105506479 on Build GRCh38
6450467 c.281G>A p.W94* Chr.9: 105506564-105506564 on Build GRCh38
1103568 c.762T>C p.I254I Chr.9: 105506574-105506574 on Build GRCh38
5557037 c.298A>T p.I100F Chr.9: 105506581-105506581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747287968 Chr.9: 105506558-105506558 on Build GRCh38
rs898080345 Chr.9: 105506499-105506499 on Build GRCh38
rs1027671562 Chr.9: 105506543-105506543 on Build GRCh38
rs780644105 Chr.9: 105506541-105506541 on Build GRCh38
rs1015590693 Chr.9: 105506469-105506469 on Build GRCh38
rs1032959117 Chr.9: 105506615-105506615 on Build GRCh38
rs758870795 Chr.9: 105506471-105506471 on Build GRCh38
rs755384711 Chr.9: 105506591-105506591 on Build GRCh38
rs765360764 Chr.9: 105506448-105506448 on Build GRCh38
rs979985044 Chr.9: 105506599-105506599 on Build GRCh38
rs994807149 Chr.9: 105506528-105506528 on Build GRCh38
rs969908894 Chr.9: 105506597-105506597 on Build GRCh38
rs76381184 Chr.9: 105506465-105506465 on Build GRCh38
rs886206834 Chr.9: 105506491-105506491 on Build GRCh38
rs991555075 Chr.9: 105506600-105506600 on Build GRCh38

View Full Product Details