Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6425765-6426030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAGLB
Entrez Gene ID
221955
Gene Name
diacylglycerol lipase beta
Gene Aliases
DAGLBETA, KCCR13L
UniGene
Hs.487498
Gene Chromosome Location
Chr.7: 6409116-6448012 on Build GRCh38
Transcripts
NM_001142936.1 NM_139179.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6235655 c.1038C>T p.H346H Chr.7: 6426006-6426006 on Build GRCh38
4626057 c.1050T>C p.H350H Chr.7: 6425994-6425994 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1027274182 Chr.7: 6425814-6425814 on Build GRCh38
rs2272156 Chr.7: 6425841-6425841 on Build GRCh38
rs114115270 Chr.7: 6425865-6425865 on Build GRCh38
rs1055146641 Chr.7: 6425958-6425958 on Build GRCh38
rs553688630 Chr.7: 6425798-6425798 on Build GRCh38
rs562063595 Chr.7: 6425899-6425899 on Build GRCh38
rs576213796 Chr.7: 6425845-6425845 on Build GRCh38
rs945481119 Chr.7: 6425902-6425902 on Build GRCh38
rs943559103 Chr.7: 6425847-6425847 on Build GRCh38
rs993497328 Chr.7: 6425914-6425914 on Build GRCh38
rs201562957 Chr.7: 6426005-6426005 on Build GRCh38
rs966708546 Chr.7: 6425843-6425843 on Build GRCh38
rs140347773 Chr.7: 6426009-6426009 on Build GRCh38
rs201366018 Chr.7: 6425970-6425970 on Build GRCh38
rs774115275 Chr.7: 6425982-6425982 on Build GRCh38
rs993199047 Chr.7: 6425997-6425997 on Build GRCh38
rs1043869066 Chr.7: 6425913-6425913 on Build GRCh38
rs951817997 Chr.7: 6426006-6426006 on Build GRCh38
rs371869467 Chr.7: 6425951-6425951 on Build GRCh38
rs199656329 Chr.7: 6425979-6425979 on Build GRCh38
rs761556520 Chr.7: 6425986-6425986 on Build GRCh38
rs568974887 Chr.7: 6425900-6425900 on Build GRCh38
rs961787488 Chr.7: 6425807-6425807 on Build GRCh38
rs760534350 Chr.7: 6426003-6426003 on Build GRCh38
rs767427403 Chr.7: 6425995-6425995 on Build GRCh38
rs976243670 Chr.7: 6425846-6425846 on Build GRCh38
rs374059982 Chr.7: 6425968-6425968 on Build GRCh38
rs778878162 Chr.7: 6425937-6425937 on Build GRCh38
rs836524 Chr.7: 6425967-6425967 on Build GRCh38
rs558723666 Chr.7: 6425833-6425833 on Build GRCh38
rs899453703 Chr.7: 6425911-6425911 on Build GRCh38
rs758374253 Chr.7: 6425955-6425955 on Build GRCh38
rs771350846 Chr.7: 6425966-6425966 on Build GRCh38
rs867768339 Chr.7: 6425842-6425842 on Build GRCh38
rs774895598 Chr.7: 6425972-6425972 on Build GRCh38
rs778105855 Chr.7: 6425965-6425965 on Build GRCh38
rs2272157 Chr.7: 6425810-6425810 on Build GRCh38
rs934313379 Chr.7: 6425828-6425829 on Build GRCh38
rs372678104 Chr.7: 6425980-6425980 on Build GRCh38

View Full Product Details