Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 95101740-95101965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf3
Entrez Gene ID
84909
Gene Name
chromosome 9 open reading frame 3
Gene Aliases
AOPEP, AP-O, APO, C90RF3, ONPEP
UniGene
Hs.434253
Gene Chromosome Location
Chr.9: 94726604-95148264 on Build GRCh38
Transcripts
XM_011519121.2 NM_000136.2 NM_001243743.1 XM_005251802.3 XM_006717001.2 XM_011518365.2 XM_011518367.2 XM_017014452.1 XM_017014453.1 XM_017014454.1

Gene Symbol
FANCC
Entrez Gene ID
2176
Gene Name
Fanconi anemia complementation group C
Gene Aliases
FA3, FAC, FACC
UniGene
Hs.494529
Gene Chromosome Location
Chr.9: 95099054-95317730 on Build GRCh38
Transcripts
XM_011519121.2 NM_000136.2 NM_001243743.1 XM_005251802.3 XM_006717001.2 XM_011518365.2 XM_011518367.2 XM_017014452.1 XM_017014453.1 XM_017014454.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
275088 c.1566C>A p.I522I Chr.9: 95101818-95101818 on Build GRCh38
753818 c.1550A>T p.E517V Chr.9: 95101834-95101834 on Build GRCh38
1111381 c.1603C>T p.R535C Chr.9: 95101781-95101781 on Build GRCh38
6958651 c.1619G>A p.S540N Chr.9: 95101765-95101765 on Build GRCh38
138720 c.65G>A p.W22* Chr.9: 95101785-95101785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766044261 Chr.9: 95101920-95101920 on Build GRCh38
rs781445050 Chr.9: 95101822-95101822 on Build GRCh38
rs761359937 Chr.9: 95101902-95101902 on Build GRCh38
rs776413570 Chr.9: 95101891-95101891 on Build GRCh38
rs990745009 Chr.9: 95101919-95101919 on Build GRCh38
rs587779901 Chr.9: 95101809-95101809 on Build GRCh38
rs201536844 Chr.9: 95101873-95101873 on Build GRCh38
rs766410371 Chr.9: 95101762-95101762 on Build GRCh38
rs587779902 Chr.9: 95101780-95101780 on Build GRCh38
rs185822330 Chr.9: 95101781-95101781 on Build GRCh38
rs780090887 Chr.9: 95101867-95101867 on Build GRCh38
rs1057516455 Chr.9: 95101785-95101785 on Build GRCh38
rs1027358273 Chr.9: 95101773-95101773 on Build GRCh38
rs587779903 Chr.9: 95101777-95101777 on Build GRCh38
rs730881727 Chr.9: 95101855-95101855 on Build GRCh38
rs748342368 Chr.9: 95101855-95101855 on Build GRCh38
rs766809608 Chr.9: 95101795-95101795 on Build GRCh38
rs764649916 Chr.9: 95101790-95101790 on Build GRCh38
rs758842354 Chr.9: 95101791-95101791 on Build GRCh38
rs1015147630 Chr.9: 95101797-95101797 on Build GRCh38
rs150020474 Chr.9: 95101824-95101824 on Build GRCh38
rs755289392 Chr.9: 95101814-95101814 on Build GRCh38
rs111916528 Chr.9: 95101933-95101933 on Build GRCh38
rs730881728 Chr.9: 95101787-95101787 on Build GRCh38
rs749319183 Chr.9: 95101844-95101844 on Build GRCh38
rs988780618 Chr.9: 95101796-95101796 on Build GRCh38
rs981705777 Chr.9: 95101836-95101836 on Build GRCh38
rs775430136 Chr.9: 95101774-95101774 on Build GRCh38
rs201379302 Chr.9: 95101840-95101840 on Build GRCh38
rs995841421 Chr.9: 95101770-95101770 on Build GRCh38
rs752855423 Chr.9: 95101823-95101823 on Build GRCh38
rs938130293 Chr.9: 95101878-95101878 on Build GRCh38
rs55939573 Chr.9: 95101789-95101789 on Build GRCh38
rs536836859 Chr.9: 95101859-95101859 on Build GRCh38
rs794726667 Chr.9: 95101828-95101829 on Build GRCh38
rs587778326 Chr.9: 95101799-95101799 on Build GRCh38
rs971409322 Chr.9: 95101811-95101811 on Build GRCh38

View Full Product Details