Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 31405460-31405731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DROSHA
Entrez Gene ID
29102
Gene Name
drosha ribonuclease III
Gene Aliases
ETOHI2, HSA242976, RANSE3L, RN3, RNASE3L, RNASEN
UniGene
Hs.97997
Gene Chromosome Location
Chr.5: 31400494-31532175 on Build GRCh38
Transcripts
NM_001100412.1 NM_013235.4 XM_005248291.3 XM_005248292.3 XM_005248293.3 XM_005248294.3 XM_017009399.1 XM_017009400.1 XM_017009401.1 XM_017009402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6293362 c.3987T>A p.L1329L Chr.5: 31405684-31405684 on Build GRCh38
3014360 c.3960G>A p.A1320A Chr.5: 31405711-31405711 on Build GRCh38
4961160 c.3973G>A p.A1325T Chr.5: 31405698-31405698 on Build GRCh38
3721572 c.3971C>G p.A1324G Chr.5: 31405700-31405700 on Build GRCh38
7301455 c.3993_3994insA p.Y1332fs*2 Chr.5: 31405677-31405678 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs75002788 Chr.5: 31405644-31405644 on Build GRCh38
rs777190304 Chr.5: 31405659-31405663 on Build GRCh38
rs922116098 Chr.5: 31405542-31405542 on Build GRCh38
rs764643911 Chr.5: 31405659-31405659 on Build GRCh38
rs567200887 Chr.5: 31405546-31405546 on Build GRCh38
rs778908885 Chr.5: 31405650-31405650 on Build GRCh38
rs777902212 Chr.5: 31405633-31405633 on Build GRCh38
rs879340504 Chr.5: 31405674-31405674 on Build GRCh38
rs768578400 Chr.5: 31405683-31405683 on Build GRCh38
rs374841209 Chr.5: 31405510-31405510 on Build GRCh38
rs534741334 Chr.5: 31405658-31405659 on Build GRCh38
rs547178328 Chr.5: 31405521-31405521 on Build GRCh38
rs867231759 Chr.5: 31405686-31405686 on Build GRCh38
rs966865209 Chr.5: 31405617-31405617 on Build GRCh38
rs549564455 Chr.5: 31405626-31405626 on Build GRCh38
rs772206875 Chr.5: 31405654-31405654 on Build GRCh38
rs757990312 Chr.5: 31405631-31405631 on Build GRCh38
rs749504913 Chr.5: 31405641-31405641 on Build GRCh38
rs557124135 Chr.5: 31405656-31405656 on Build GRCh38
rs569320628 Chr.5: 31405632-31405632 on Build GRCh38
rs776295904 Chr.5: 31405687-31405687 on Build GRCh38
rs746746592 Chr.5: 31405669-31405669 on Build GRCh38
rs898901219 Chr.5: 31405695-31405695 on Build GRCh38
rs913315166 Chr.5: 31405512-31405512 on Build GRCh38
rs745786738 Chr.5: 31405652-31405652 on Build GRCh38
rs529515985 Chr.5: 31405583-31405583 on Build GRCh38
rs978937669 Chr.5: 31405527-31405527 on Build GRCh38

View Full Product Details