Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 33114430-33114661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTAP
Entrez Gene ID
10491
Gene Name
cartilage associated protein
Gene Aliases
CASP, LEPREL3, OI7, P3H5
UniGene
Hs.517888
Gene Chromosome Location
Chr.3: 33113958-33147773 on Build GRCh38
Transcripts
NM_006371.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
730308 c.443A>G p.Y148C Chr.3: 33114520-33114520 on Build GRCh38
3993032 c.439C>T p.P147S Chr.3: 33114516-33114516 on Build GRCh38
5974736 c.456G>C p.Q152H Chr.3: 33114533-33114533 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1006755891 Chr.3: 33114613-33114613 on Build GRCh38
rs931952062 Chr.3: 33114469-33114469 on Build GRCh38
rs1057480002 Chr.3: 33114562-33114562 on Build GRCh38
rs921862940 Chr.3: 33114457-33114457 on Build GRCh38
rs778764392 Chr.3: 33114464-33114464 on Build GRCh38
rs774287562 Chr.3: 33114564-33114564 on Build GRCh38
rs531119589 Chr.3: 33114536-33114536 on Build GRCh38
rs762232251 Chr.3: 33114494-33114494 on Build GRCh38
rs972668240 Chr.3: 33114521-33114521 on Build GRCh38
rs137853943 Chr.3: 33114550-33114550 on Build GRCh38
rs759716355 Chr.3: 33114566-33114566 on Build GRCh38
rs17850371 Chr.3: 33114488-33114488 on Build GRCh38
rs773473218 Chr.3: 33114497-33114497 on Build GRCh38
rs1028990727 Chr.3: 33114589-33114589 on Build GRCh38
rs776984474 Chr.3: 33114489-33114489 on Build GRCh38
rs935063560 Chr.3: 33114498-33114498 on Build GRCh38
rs953400115 Chr.3: 33114608-33114608 on Build GRCh38
rs969878155 Chr.3: 33114465-33114465 on Build GRCh38
rs747496610 Chr.3: 33114486-33114486 on Build GRCh38
rs949350747 Chr.3: 33114586-33114586 on Build GRCh38
rs137853942 Chr.3: 33114546-33114546 on Build GRCh38
rs911685787 Chr.3: 33114630-33114630 on Build GRCh38
rs771525192 Chr.3: 33114479-33114479 on Build GRCh38
rs979623061 Chr.3: 33114559-33114559 on Build GRCh38
rs755889115 Chr.3: 33114449-33114449 on Build GRCh38
rs757163727 Chr.3: 33114516-33114516 on Build GRCh38
rs776028033 Chr.3: 33114483-33114483 on Build GRCh38
rs896763854 Chr.3: 33114574-33114574 on Build GRCh38
rs202118861 Chr.3: 33114528-33114528 on Build GRCh38
rs549296015 Chr.3: 33114552-33114552 on Build GRCh38
rs770975997 Chr.3: 33114561-33114561 on Build GRCh38
rs763097630 Chr.3: 33114557-33114557 on Build GRCh38
rs1045050044 Chr.3: 33114597-33114597 on Build GRCh38
rs201564256 Chr.3: 33114523-33114523 on Build GRCh38
rs987708122 Chr.3: 33114476-33114476 on Build GRCh38
rs750198183 Chr.3: 33114560-33114560 on Build GRCh38
rs1016748122 Chr.3: 33114625-33114625 on Build GRCh38
rs962653319 Chr.3: 33114518-33114518 on Build GRCh38
rs777442040 Chr.3: 33114450-33114450 on Build GRCh38
rs753670727 Chr.3: 33114512-33114512 on Build GRCh38
rs956567710 Chr.3: 33114631-33114631 on Build GRCh38
rs963317151 Chr.3: 33114576-33114576 on Build GRCh38
rs1052610154 Chr.3: 33114572-33114572 on Build GRCh38
rs779447329 Chr.3: 33114533-33114533 on Build GRCh38
rs756961672 Chr.3: 33114453-33114453 on Build GRCh38
rs137853941 Chr.3: 33114481-33114481 on Build GRCh38
rs766390490 Chr.3: 33114506-33114506 on Build GRCh38
rs745346112 Chr.3: 33114475-33114475 on Build GRCh38
rs532846004 Chr.3: 33114507-33114507 on Build GRCh38
rs765434106 Chr.3: 33114496-33114496 on Build GRCh38
rs72659359 Chr.3: 33114549-33114549 on Build GRCh38
rs752605909 Chr.3: 33114448-33114448 on Build GRCh38
rs905266269 Chr.3: 33114598-33114598 on Build GRCh38
rs748878855 Chr.3: 33114452-33114452 on Build GRCh38
rs763913403 Chr.3: 33114585-33114585 on Build GRCh38
rs773371198 Chr.3: 33114555-33114555 on Build GRCh38
rs770173723 Chr.3: 33114553-33114553 on Build GRCh38
rs879273215 Chr.3: 33114503-33114503 on Build GRCh38
rs762979572 Chr.3: 33114505-33114505 on Build GRCh38

View Full Product Details