Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61415589-61415844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF7
Entrez Gene ID
79869
Gene Name
cleavage and polyadenylation specific factor 7
Gene Aliases
CFIm59
UniGene
Hs.718984
Gene Chromosome Location
Chr.11: 61402648-61429992 on Build GRCh38
Transcripts
NM_001136040.2 NM_001142565.1 NM_024811.3 XM_005274298.4 XM_005274299.4 XM_005274303.4 XM_011545257.2 XM_011545258.2 XM_011545259.2 XM_011545260.2 XM_011545261.2 XM_011545262.2 XM_011545263.2 XM_017018345.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4198528 c.1104G>A p.S368S Chr.11: 61415775-61415775 on Build GRCh38
2037501 c.975G>A p.S325S Chr.11: 61415775-61415775 on Build GRCh38
4866814 c.1213+1G>A p.? Chr.11: 61415665-61415665 on Build GRCh38
929461 c.1084+1G>A p.? Chr.11: 61415665-61415665 on Build GRCh38
1676084 c.1027C>T p.R343* Chr.11: 61415723-61415723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771392757 Chr.11: 61415788-61415788 on Build GRCh38
rs370996325 Chr.11: 61415710-61415711 on Build GRCh38
rs775722687 Chr.11: 61415708-61415708 on Build GRCh38
rs770308764 Chr.11: 61415645-61415645 on Build GRCh38
rs961372134 Chr.11: 61415637-61415637 on Build GRCh38
rs749745722 Chr.11: 61415776-61415776 on Build GRCh38
rs754453926 Chr.11: 61415773-61415773 on Build GRCh38
rs1064377 Chr.11: 61415699-61415699 on Build GRCh38
rs773669592 Chr.11: 61415649-61415649 on Build GRCh38
rs781576103 Chr.11: 61415632-61415634 on Build GRCh38
rs977204304 Chr.11: 61415750-61415750 on Build GRCh38
rs374286273 Chr.11: 61415615-61415615 on Build GRCh38
rs535339368 Chr.11: 61415759-61415759 on Build GRCh38
rs200619739 Chr.11: 61415667-61415667 on Build GRCh38
rs778301230 Chr.11: 61415775-61415775 on Build GRCh38
rs749703368 Chr.11: 61415654-61415654 on Build GRCh38
rs746465965 Chr.11: 61415642-61415642 on Build GRCh38
rs757996507 Chr.11: 61415629-61415629 on Build GRCh38
rs746183295 Chr.11: 61415791-61415791 on Build GRCh38
rs758062445 Chr.11: 61415756-61415756 on Build GRCh38
rs370060256 Chr.11: 61415702-61415702 on Build GRCh38
rs375587977 Chr.11: 61415812-61415812 on Build GRCh38
rs377249463 Chr.11: 61415684-61415684 on Build GRCh38
rs774581220 Chr.11: 61415789-61415789 on Build GRCh38
rs764941275 Chr.11: 61415752-61415752 on Build GRCh38
rs200649476 Chr.11: 61415801-61415801 on Build GRCh38
rs746553918 Chr.11: 61415767-61415767 on Build GRCh38
rs759671978 Chr.11: 61415697-61415697 on Build GRCh38
rs369255329 Chr.11: 61415754-61415754 on Build GRCh38
rs779546119 Chr.11: 61415630-61415630 on Build GRCh38
rs149166727 Chr.11: 61415745-61415745 on Build GRCh38
rs772442908 Chr.11: 61415792-61415792 on Build GRCh38
rs559063740 Chr.11: 61415748-61415748 on Build GRCh38
rs373141451 Chr.11: 61415730-61415730 on Build GRCh38
rs760785328 Chr.11: 61415721-61415721 on Build GRCh38
rs370561539 Chr.11: 61415704-61415704 on Build GRCh38
rs775810707 Chr.11: 61415799-61415799 on Build GRCh38

View Full Product Details