Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 200873206-200873451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR25
Entrez Gene ID
2848
Gene Name
G protein-coupled receptor 25
Gene Aliases
-
UniGene
Hs.534316
Gene Chromosome Location
Chr.1: 200872955-200874178 on Build GRCh38
Transcripts
NM_005298.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6508455 c.289G>A p.A97T Chr.1: 200873326-200873326 on Build GRCh38
358313 c.388C>G p.L130V Chr.1: 200873425-200873425 on Build GRCh38
5082105 c.297G>A p.A99A Chr.1: 200873334-200873334 on Build GRCh38
6045332 c.352G>A p.A118T Chr.1: 200873389-200873389 on Build GRCh38
1601499 c.273G>C p.T91T Chr.1: 200873310-200873310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs865905946 Chr.1: 200873404-200873404 on Build GRCh38
rs573276776 Chr.1: 200873358-200873358 on Build GRCh38
rs765139398 Chr.1: 200873268-200873268 on Build GRCh38
rs956610201 Chr.1: 200873367-200873367 on Build GRCh38
rs762581931 Chr.1: 200873331-200873331 on Build GRCh38
rs774361103 Chr.1: 200873238-200873238 on Build GRCh38
rs866466582 Chr.1: 200873387-200873387 on Build GRCh38
rs1037035534 Chr.1: 200873399-200873399 on Build GRCh38
rs903882947 Chr.1: 200873293-200873293 on Build GRCh38
rs775285159 Chr.1: 200873429-200873429 on Build GRCh38
rs756415380 Chr.1: 200873426-200873426 on Build GRCh38
rs768767160 Chr.1: 200873233-200873233 on Build GRCh38
rs776741589 Chr.1: 200873257-200873257 on Build GRCh38
rs868812839 Chr.1: 200873321-200873321 on Build GRCh38
rs868429254 Chr.1: 200873389-200873389 on Build GRCh38
rs866759371 Chr.1: 200873320-200873320 on Build GRCh38
rs775197755 Chr.1: 200873271-200873271 on Build GRCh38
rs751075873 Chr.1: 200873361-200873361 on Build GRCh38
rs931396742 Chr.1: 200873432-200873432 on Build GRCh38
rs536263225 Chr.1: 200873376-200873376 on Build GRCh38
rs773720673 Chr.1: 200873402-200873414 on Build GRCh38
rs940033627 Chr.1: 200873355-200873355 on Build GRCh38
rs912840423 Chr.1: 200873302-200873302 on Build GRCh38
rs750936473 Chr.1: 200873418-200873418 on Build GRCh38
rs759339832 Chr.1: 200873265-200873265 on Build GRCh38
rs61731829 Chr.1: 200873415-200873415 on Build GRCh38
rs1018351275 Chr.1: 200873246-200873246 on Build GRCh38
rs749484042 Chr.1: 200873232-200873232 on Build GRCh38
rs770943664 Chr.1: 200873249-200873249 on Build GRCh38
rs767975347 Chr.1: 200873398-200873398 on Build GRCh38
rs919971689 Chr.1: 200873422-200873422 on Build GRCh38

View Full Product Details