Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16478940-16479213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALR3
Entrez Gene ID
125972
Gene Name
calreticulin 3
Gene Aliases
CMH19, CRT2, CT93
UniGene
Hs.304020
Gene Chromosome Location
Chr.19: 16479057-16496192 on Build GRCh38
Transcripts
NM_145046.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5601185 c.1097G>A p.G366E Chr.19: 16479189-16479189 on Build GRCh38
7263373 c.1125C>T p.F375F Chr.19: 16479161-16479161 on Build GRCh38
3422516 c.1131A>C p.Q377H Chr.19: 16479155-16479155 on Build GRCh38
5916695 c.1142G>A p.R381K Chr.19: 16479144-16479144 on Build GRCh38
3284619 c.1114G>A p.E372K Chr.19: 16479172-16479172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896891002 Chr.19: 16479053-16479053 on Build GRCh38
rs1019355219 Chr.19: 16479002-16479002 on Build GRCh38
rs963841276 Chr.19: 16479038-16479038 on Build GRCh38
rs375160369 Chr.19: 16479171-16479171 on Build GRCh38
rs996666654 Chr.19: 16479058-16479058 on Build GRCh38
rs372514243 Chr.19: 16479157-16479157 on Build GRCh38
rs968684576 Chr.19: 16479075-16479076 on Build GRCh38
rs768736111 Chr.19: 16479173-16479173 on Build GRCh38
rs545887392 Chr.19: 16479076-16479076 on Build GRCh38
rs118051449 Chr.19: 16478996-16478996 on Build GRCh38
rs200844516 Chr.19: 16479191-16479191 on Build GRCh38
rs368434498 Chr.19: 16479139-16479139 on Build GRCh38
rs145534922 Chr.19: 16479110-16479110 on Build GRCh38
rs746938291 Chr.19: 16479148-16479148 on Build GRCh38
rs768015132 Chr.19: 16479192-16479192 on Build GRCh38
rs545737929 Chr.19: 16479062-16479062 on Build GRCh38
rs778754506 Chr.19: 16479124-16479124 on Build GRCh38
rs774421614 Chr.19: 16479176-16479176 on Build GRCh38
rs140440387 Chr.19: 16479172-16479172 on Build GRCh38
rs762323585 Chr.19: 16479087-16479087 on Build GRCh38
rs750756265 Chr.19: 16479111-16479111 on Build GRCh38
rs200723554 Chr.19: 16478975-16478975 on Build GRCh38
rs777411023 Chr.19: 16479140-16479140 on Build GRCh38
rs1037886844 Chr.19: 16479177-16479177 on Build GRCh38
rs943874529 Chr.19: 16479004-16479004 on Build GRCh38
rs752364126 Chr.19: 16479136-16479136 on Build GRCh38
rs1036907567 Chr.19: 16479008-16479008 on Build GRCh38
rs754605848 Chr.19: 16479113-16479113 on Build GRCh38
rs767040811 Chr.19: 16479155-16479155 on Build GRCh38

View Full Product Details