Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102836948-102837221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP17A1
Entrez Gene ID
1586
Gene Name
cytochrome P450 family 17 subfamily A member 1
Gene Aliases
CPT7, CYP17, P450C17, S17AH
UniGene
Hs.438016
Gene Chromosome Location
Chr.10: 102830531-102837533 on Build GRCh38
Transcripts
NM_000102.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1718558 c.273G>A p.K91K Chr.10: 102837089-102837089 on Build GRCh38
465209 c.251A>T p.E84V Chr.10: 102837111-102837111 on Build GRCh38
6908025 c.163A>G p.K55E Chr.10: 102837199-102837199 on Build GRCh38
1702964 c.257T>C p.L86P Chr.10: 102837105-102837105 on Build GRCh38
3433805 c.250G>A p.E84K Chr.10: 102837112-102837112 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs104894153 Chr.10: 102837075-102837075 on Build GRCh38
rs61754265 Chr.10: 102837133-102837133 on Build GRCh38
rs568494540 Chr.10: 102837031-102837031 on Build GRCh38
rs752831246 Chr.10: 102837058-102837058 on Build GRCh38
rs104894146 Chr.10: 102837084-102837084 on Build GRCh38
rs776954575 Chr.10: 102837048-102837048 on Build GRCh38
rs757599828 Chr.10: 102837069-102837069 on Build GRCh38
rs751850976 Chr.10: 102837065-102837065 on Build GRCh38
rs965463189 Chr.10: 102837038-102837038 on Build GRCh38
rs1034193804 Chr.10: 102836996-102836996 on Build GRCh38
rs750853745 Chr.10: 102837163-102837163 on Build GRCh38
rs187269884 Chr.10: 102836987-102836987 on Build GRCh38
rs759770001 Chr.10: 102837119-102837119 on Build GRCh38
rs376074317 Chr.10: 102837162-102837162 on Build GRCh38
rs781079819 Chr.10: 102837143-102837143 on Build GRCh38
rs1011510315 Chr.10: 102837015-102837015 on Build GRCh38
rs974635568 Chr.10: 102837125-102837125 on Build GRCh38
rs778389140 Chr.10: 102837093-102837093 on Build GRCh38
rs535575638 Chr.10: 102837086-102837086 on Build GRCh38
rs6163 Chr.10: 102837167-102837167 on Build GRCh38
rs770800801 Chr.10: 102837110-102837110 on Build GRCh38
rs538423333 Chr.10: 102837009-102837009 on Build GRCh38
rs776580666 Chr.10: 102837114-102837114 on Build GRCh38
rs908864807 Chr.10: 102837154-102837154 on Build GRCh38
rs758829043 Chr.10: 102837059-102837059 on Build GRCh38
rs765350010 Chr.10: 102837052-102837052 on Build GRCh38
rs894343961 Chr.10: 102837002-102837002 on Build GRCh38
rs931528592 Chr.10: 102837137-102837137 on Build GRCh38
rs763145133 Chr.10: 102837134-102837134 on Build GRCh38
rs754611524 Chr.10: 102837164-102837164 on Build GRCh38
rs760564520 Chr.10: 102837035-102837035 on Build GRCh38
rs370973897 Chr.10: 102837127-102837127 on Build GRCh38
rs111865215 Chr.10: 102837044-102837044 on Build GRCh38
rs571102919 Chr.10: 102836990-102836990 on Build GRCh38
rs61754264 Chr.10: 102837182-102837182 on Build GRCh38
rs747750444 Chr.10: 102837101-102837101 on Build GRCh38
rs764723654 Chr.10: 102837063-102837063 on Build GRCh38
rs967398653 Chr.10: 102837026-102837026 on Build GRCh38
rs761588720 Chr.10: 102837019-102837019 on Build GRCh38
rs104894138 Chr.10: 102837076-102837076 on Build GRCh38
rs202192337 Chr.10: 102837051-102837051 on Build GRCh38
rs771880642 Chr.10: 102837102-102837102 on Build GRCh38
rs553716444 Chr.10: 102837104-102837104 on Build GRCh38
rs777171001 Chr.10: 102837178-102837178 on Build GRCh38
rs906926519 Chr.10: 102837168-102837168 on Build GRCh38
rs1022013950 Chr.10: 102837024-102837024 on Build GRCh38
rs45473095 Chr.10: 102837027-102837027 on Build GRCh38
rs1002694576 Chr.10: 102836994-102836994 on Build GRCh38
rs766680191 Chr.10: 102837046-102837046 on Build GRCh38
rs370791765 Chr.10: 102837177-102837177 on Build GRCh38
rs752052614 Chr.10: 102837160-102837160 on Build GRCh38

View Full Product Details