Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151505715-151505966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAT2
Entrez Gene ID
2196
Gene Name
FAT atypical cadherin 2
Gene Aliases
CDHF8, CDHR9, HFAT2, MEGF1
UniGene
Hs.591255
Gene Chromosome Location
Chr.5: 151504092-151594822 on Build GRCh38
Transcripts
NM_001447.2 XM_006714761.3 XM_011537600.2 XM_011537603.2 XM_017009224.1 XM_017009225.1 XM_011537595.2 XM_017009217.1 XM_017009219.1

Gene Symbol
SLC36A1
Entrez Gene ID
206358
Gene Name
solute carrier family 36 member 1
Gene Aliases
Dct1, LYAAT1, PAT1, TRAMD3
UniGene
Hs.269004
Gene Chromosome Location
Chr.5: 151411344-151556318 on Build GRCh38
Transcripts
NM_001447.2 XM_006714761.3 XM_011537600.2 XM_011537603.2 XM_017009224.1 XM_017009225.1 XM_011537595.2 XM_017009217.1 XM_017009219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6660876 c.12649C>A p.P4217T Chr.5: 151505966-151505966 on Build GRCh38
6546814 c.12684C>A p.F4228L Chr.5: 151505931-151505931 on Build GRCh38
6660880 c.12779G>A p.R4260Q Chr.5: 151505836-151505836 on Build GRCh38
4564558 c.12723_12724CC>TT p.R4242C Chr.5: 151505891-151505892 on Build GRCh38
6276637 c.12702C>A p.N4234K Chr.5: 151505913-151505913 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767594543 Chr.5: 151505912-151505912 on Build GRCh38
rs745551428 Chr.5: 151505809-151505809 on Build GRCh38
rs756083474 Chr.5: 151505783-151505783 on Build GRCh38
rs199734683 Chr.5: 151505738-151505738 on Build GRCh38
rs771437559 Chr.5: 151505815-151505815 on Build GRCh38
rs756029200 Chr.5: 151505837-151505837 on Build GRCh38
rs200478785 Chr.5: 151505808-151505808 on Build GRCh38
rs761235764 Chr.5: 151505820-151505820 on Build GRCh38
rs752588731 Chr.5: 151505778-151505778 on Build GRCh38
rs753672644 Chr.5: 151505919-151505919 on Build GRCh38
rs376159254 Chr.5: 151505865-151505865 on Build GRCh38
rs770756961 Chr.5: 151505797-151505797 on Build GRCh38
rs753815387 Chr.5: 151505845-151505845 on Build GRCh38
rs779690051 Chr.5: 151505886-151505886 on Build GRCh38
rs375900487 Chr.5: 151505908-151505908 on Build GRCh38
rs750656516 Chr.5: 151505816-151505817 on Build GRCh38
rs372212910 Chr.5: 151505825-151505825 on Build GRCh38
rs764912855 Chr.5: 151505926-151505926 on Build GRCh38
rs570779199 Chr.5: 151505909-151505909 on Build GRCh38
rs771799746 Chr.5: 151505858-151505858 on Build GRCh38
rs765038227 Chr.5: 151505852-151505852 on Build GRCh38
rs762151076 Chr.5: 151505827-151505827 on Build GRCh38
rs751686564 Chr.5: 151505836-151505836 on Build GRCh38
rs529089589 Chr.5: 151505816-151505816 on Build GRCh38
rs780204714 Chr.5: 151505777-151505777 on Build GRCh38
rs765639933 Chr.5: 151505828-151505828 on Build GRCh38
rs765435793 Chr.5: 151505765-151505765 on Build GRCh38
rs769281607 Chr.5: 151505891-151505891 on Build GRCh38
rs545506007 Chr.5: 151505739-151505739 on Build GRCh38
rs1045093533 Chr.5: 151505904-151505904 on Build GRCh38
rs144888662 Chr.5: 151505890-151505890 on Build GRCh38
rs754295945 Chr.5: 151505762-151505762 on Build GRCh38
rs763266607 Chr.5: 151505831-151505831 on Build GRCh38
rs756948000 Chr.5: 151505849-151505849 on Build GRCh38
rs750561477 Chr.5: 151505830-151505830 on Build GRCh38
rs773895524 Chr.5: 151505746-151505746 on Build GRCh38
rs557699400 Chr.5: 151505748-151505748 on Build GRCh38
rs975144874 Chr.5: 151505784-151505786 on Build GRCh38
rs552300011 Chr.5: 151505903-151505903 on Build GRCh38
rs199880355 Chr.5: 151505892-151505892 on Build GRCh38
rs770014066 Chr.5: 151505896-151505896 on Build GRCh38
rs770608155 Chr.5: 151505800-151505800 on Build GRCh38
rs372063434 Chr.5: 151505799-151505799 on Build GRCh38
rs762403470 Chr.5: 151505895-151505895 on Build GRCh38
rs1022132958 Chr.5: 151505753-151505753 on Build GRCh38
rs766412867 Chr.5: 151505834-151505834 on Build GRCh38
rs758351608 Chr.5: 151505771-151505771 on Build GRCh38
rs750285304 Chr.5: 151505931-151505931 on Build GRCh38
rs760521076 Chr.5: 151505743-151505743 on Build GRCh38
rs371252623 Chr.5: 151505749-151505749 on Build GRCh38
rs750513372 Chr.5: 151505770-151505770 on Build GRCh38
rs149865780 Chr.5: 151505761-151505761 on Build GRCh38
rs375536782 Chr.5: 151505838-151505838 on Build GRCh38
rs199540723 Chr.5: 151505752-151505752 on Build GRCh38
rs757254130 Chr.5: 151505920-151505920 on Build GRCh38
rs200172237 Chr.5: 151505851-151505851 on Build GRCh38
rs967940329 Chr.5: 151505754-151505754 on Build GRCh38

View Full Product Details