Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 42628967-42629214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHR
Entrez Gene ID
2690
Gene Name
growth hormone receptor
Gene Aliases
GHBP, GHIP
UniGene
Hs.125180
Gene Chromosome Location
Chr.5: 42423775-42721878 on Build GRCh38
Transcripts
NM_000163.4 NM_001242399.2 NM_001242400.2 NM_001242401.3 NM_001242402.2 NM_001242403.2 NM_001242404.2 NM_001242405.2 NM_001242406.2 NM_001242460.1 NM_001242462.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6992733 c.81T>A p.A27A Chr.5: 42629048-42629048 on Build GRCh38
3855333 c.83T>A p.I28N Chr.5: 42629050-42629050 on Build GRCh38
4954331 c.82A>G p.I28V Chr.5: 42629049-42629049 on Build GRCh38
3616669 c.132G>A p.K44K Chr.5: 42629099-42629099 on Build GRCh38
7369424 c.74C>G p.T25R Chr.5: 42629041-42629041 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761015692 Chr.5: 42629094-42629094 on Build GRCh38
rs377297987 Chr.5: 42629065-42629065 on Build GRCh38
rs757860934 Chr.5: 42629037-42629037 on Build GRCh38
rs756925109 Chr.5: 42629043-42629043 on Build GRCh38
rs558277834 Chr.5: 42629174-42629174 on Build GRCh38
rs776748737 Chr.5: 42629051-42629051 on Build GRCh38
rs760051357 Chr.5: 42629134-42629134 on Build GRCh38
rs781185088 Chr.5: 42629048-42629048 on Build GRCh38
rs752312134 Chr.5: 42629035-42629035 on Build GRCh38
rs772258414 Chr.5: 42629091-42629091 on Build GRCh38
rs373055838 Chr.5: 42629040-42629040 on Build GRCh38
rs181298988 Chr.5: 42629086-42629086 on Build GRCh38
rs777009328 Chr.5: 42629121-42629121 on Build GRCh38
rs1014385043 Chr.5: 42629071-42629071 on Build GRCh38
rs76183160 Chr.5: 42629066-42629066 on Build GRCh38
rs371117905 Chr.5: 42629104-42629104 on Build GRCh38
rs763594388 Chr.5: 42629039-42629039 on Build GRCh38
rs121909370 Chr.5: 42629069-42629069 on Build GRCh38
rs369825370 Chr.5: 42629002-42629002 on Build GRCh38
rs143287692 Chr.5: 42629049-42629049 on Build GRCh38
rs756184879 Chr.5: 42629050-42629050 on Build GRCh38
rs773634630 Chr.5: 42629093-42629093 on Build GRCh38
rs759746207 Chr.5: 42629062-42629062 on Build GRCh38
rs774448311 Chr.5: 42629085-42629085 on Build GRCh38
rs779840197 Chr.5: 42629059-42629059 on Build GRCh38
rs904748525 Chr.5: 42629045-42629045 on Build GRCh38

View Full Product Details