Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 28785952-28786177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC117
Entrez Gene ID
150275
Gene Name
coiled-coil domain containing 117
Gene Aliases
dJ366L4.1
UniGene
Hs.406460
Gene Chromosome Location
Chr.22: 28772674-28789301 on Build GRCh38
Transcripts
NM_001284263.1 NM_001284264.1 NM_001284265.1 NM_173510.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6575528 c.642C>T p.L214L Chr.22: 28786128-28786128 on Build GRCh38
127552 c.630C>A p.L210L Chr.22: 28786116-28786116 on Build GRCh38
3701727 c.603-6T>C p.? Chr.22: 28786083-28786083 on Build GRCh38
4103249 c.607C>T p.R203C Chr.22: 28786093-28786093 on Build GRCh38
3553062 c.652C>T p.L218F Chr.22: 28786138-28786138 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766241307 Chr.22: 28786056-28786056 on Build GRCh38
rs760794325 Chr.22: 28786007-28786007 on Build GRCh38
rs767124392 Chr.22: 28786045-28786045 on Build GRCh38
rs972531837 Chr.22: 28786027-28786027 on Build GRCh38
rs752519765 Chr.22: 28786078-28786078 on Build GRCh38
rs765180063 Chr.22: 28786068-28786068 on Build GRCh38
rs754335519 Chr.22: 28786094-28786094 on Build GRCh38
rs780433074 Chr.22: 28786099-28786099 on Build GRCh38
rs769031781 Chr.22: 28786102-28786102 on Build GRCh38
rs938878113 Chr.22: 28786115-28786115 on Build GRCh38
rs749876500 Chr.22: 28786100-28786100 on Build GRCh38
rs754865644 Chr.22: 28786066-28786066 on Build GRCh38
rs781525792 Chr.22: 28786088-28786088 on Build GRCh38
rs760107421 Chr.22: 28786137-28786137 on Build GRCh38
rs150710057 Chr.22: 28785981-28785981 on Build GRCh38
rs748468959 Chr.22: 28786116-28786116 on Build GRCh38
rs771731132 Chr.22: 28786121-28786121 on Build GRCh38
rs772789093 Chr.22: 28786131-28786131 on Build GRCh38
rs749963124 Chr.22: 28786046-28786046 on Build GRCh38
rs17514713 Chr.22: 28786082-28786082 on Build GRCh38
rs745967615 Chr.22: 28786093-28786093 on Build GRCh38
rs774735302 Chr.22: 28786111-28786111 on Build GRCh38
rs373921809 Chr.22: 28786053-28786053 on Build GRCh38
rs908666795 Chr.22: 28786150-28786150 on Build GRCh38
rs753741058 Chr.22: 28786062-28786062 on Build GRCh38
rs975072274 Chr.22: 28786085-28786085 on Build GRCh38

View Full Product Details