Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 36540294-36540547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOAH
Entrez Gene ID
313
Gene Name
acyloxyacyl hydrolase
Gene Aliases
-
UniGene
Hs.488007
Gene Chromosome Location
Chr.7: 36512042-36724549 on Build GRCh38
Transcripts
NM_001177506.1 NM_001177507.1 NM_001637.3 XM_011515333.2 XM_011515334.2 XM_011515335.2 XM_011515336.2 XM_011515338.2 XM_011515339.2 XM_011515340.2 XM_011515341.2 XM_011515342.2 XM_017012102.1 XM_017012103.1 XM_017012104.1 XM_017012105.1 XM_017012106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
254534 c.1237G>A p.V413I Chr.7: 36540388-36540388 on Build GRCh38
6319711 c.1204C>A p.H402N Chr.7: 36540421-36540421 on Build GRCh38
3880818 c.1182C>T p.S394S Chr.7: 36540443-36540443 on Build GRCh38
4662176 c.1233C>T p.S411S Chr.7: 36540392-36540392 on Build GRCh38
6318419 c.1218C>A p.H406Q Chr.7: 36540407-36540407 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780318766 Chr.7: 36540404-36540404 on Build GRCh38
rs772029792 Chr.7: 36540332-36540332 on Build GRCh38
rs759056561 Chr.7: 36540450-36540450 on Build GRCh38
rs200437512 Chr.7: 36540486-36540486 on Build GRCh38
rs149685289 Chr.7: 36540374-36540374 on Build GRCh38
rs866303470 Chr.7: 36540437-36540437 on Build GRCh38
rs754260311 Chr.7: 36540500-36540500 on Build GRCh38
rs375637039 Chr.7: 36540368-36540368 on Build GRCh38
rs752785484 Chr.7: 36540496-36540498 on Build GRCh38
rs766043790 Chr.7: 36540349-36540349 on Build GRCh38
rs765010456 Chr.7: 36540466-36540466 on Build GRCh38
rs140099559 Chr.7: 36540333-36540333 on Build GRCh38
rs778926511 Chr.7: 36540427-36540427 on Build GRCh38
rs375325817 Chr.7: 36540498-36540498 on Build GRCh38
rs779122742 Chr.7: 36540508-36540508 on Build GRCh38
rs765382203 Chr.7: 36540426-36540426 on Build GRCh38
rs756492790 Chr.7: 36540400-36540400 on Build GRCh38
rs773194603 Chr.7: 36540435-36540435 on Build GRCh38
rs752184445 Chr.7: 36540467-36540467 on Build GRCh38
rs541564182 Chr.7: 36540431-36540431 on Build GRCh38
rs145831683 Chr.7: 36540440-36540440 on Build GRCh38
rs749523052 Chr.7: 36540406-36540406 on Build GRCh38
rs953251530 Chr.7: 36540487-36540487 on Build GRCh38
rs760403749 Chr.7: 36540339-36540339 on Build GRCh38
rs748233622 Chr.7: 36540428-36540428 on Build GRCh38
rs757795394 Chr.7: 36540379-36540379 on Build GRCh38
rs760519168 Chr.7: 36540439-36540439 on Build GRCh38
rs866563358 Chr.7: 36540491-36540491 on Build GRCh38
rs768945280 Chr.7: 36540407-36540407 on Build GRCh38
rs762564395 Chr.7: 36540476-36540476 on Build GRCh38
rs929014710 Chr.7: 36540457-36540457 on Build GRCh38
rs761689958 Chr.7: 36540329-36540329 on Build GRCh38
rs377250798 Chr.7: 36540481-36540481 on Build GRCh38
rs376436261 Chr.7: 36540370-36540370 on Build GRCh38
rs917686546 Chr.7: 36540446-36540446 on Build GRCh38
rs776320571 Chr.7: 36540441-36540441 on Build GRCh38
rs149792324 Chr.7: 36540380-36540380 on Build GRCh38
rs117556863 Chr.7: 36540505-36540505 on Build GRCh38
rs550416911 Chr.7: 36540360-36540360 on Build GRCh38

View Full Product Details