Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 15959722-15959993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DERA
Entrez Gene ID
51071
Gene Name
deoxyribose-phosphate aldolase
Gene Aliases
CGI-26, DEOC
UniGene
Hs.39429
Gene Chromosome Location
Chr.12: 15911172-16037381 on Build GRCh38
Transcripts
NM_001300779.1 NM_015954.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5374750 c.339C>A p.L113L Chr.12: 15959890-15959890 on Build GRCh38
5700358 c.316G>A p.V106M Chr.12: 15959867-15959867 on Build GRCh38
4677108 c.339_340delCA p.K114fs*5 Chr.12: 15959890-15959891 on Build GRCh38
5043760 c.333A>C p.K111N Chr.12: 15959884-15959884 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs139201548 Chr.12: 15959795-15959795 on Build GRCh38
rs762295838 Chr.12: 15959877-15959877 on Build GRCh38
rs777413520 Chr.12: 15959867-15959867 on Build GRCh38
rs775023184 Chr.12: 15959925-15959925 on Build GRCh38
rs371196959 Chr.12: 15959860-15959860 on Build GRCh38
rs375747326 Chr.12: 15959864-15959864 on Build GRCh38
rs79188698 Chr.12: 15959837-15959837 on Build GRCh38
rs977558014 Chr.12: 15959838-15959838 on Build GRCh38
rs546858128 Chr.12: 15959845-15959845 on Build GRCh38
rs187869190 Chr.12: 15959958-15959958 on Build GRCh38
rs912276777 Chr.12: 15959804-15959804 on Build GRCh38
rs1012130774 Chr.12: 15959763-15959763 on Build GRCh38
rs763759272 Chr.12: 15959918-15959918 on Build GRCh38
rs768740450 Chr.12: 15959871-15959871 on Build GRCh38
rs867076586 Chr.12: 15959903-15959903 on Build GRCh38
rs764932805 Chr.12: 15959894-15959894 on Build GRCh38
rs368204530 Chr.12: 15959902-15959902 on Build GRCh38
rs769952797 Chr.12: 15959846-15959846 on Build GRCh38
rs759562150 Chr.12: 15959858-15959861 on Build GRCh38
rs974043634 Chr.12: 15959755-15959755 on Build GRCh38
rs752212999 Chr.12: 15959779-15959779 on Build GRCh38
rs1056228522 Chr.12: 15959931-15959931 on Build GRCh38
rs112037247 Chr.12: 15959762-15959762 on Build GRCh38
rs530508622 Chr.12: 15959758-15959758 on Build GRCh38
rs749320478 Chr.12: 15959862-15959862 on Build GRCh38
rs773500761 Chr.12: 15959885-15959885 on Build GRCh38
rs201650092 Chr.12: 15959947-15959947 on Build GRCh38
rs115629535 Chr.12: 15959820-15959820 on Build GRCh38
rs966073823 Chr.12: 15959830-15959830 on Build GRCh38
rs763890726 Chr.12: 15959808-15959808 on Build GRCh38
rs200811658 Chr.12: 15959907-15959907 on Build GRCh38
rs921899789 Chr.12: 15959843-15959843 on Build GRCh38
rs774774116 Chr.12: 15959872-15959872 on Build GRCh38
rs59802633 Chr.12: 15959964-15959964 on Build GRCh38
rs1022249424 Chr.12: 15959776-15959776 on Build GRCh38
rs1031484026 Chr.12: 15959821-15959821 on Build GRCh38
rs370058878 Chr.12: 15959784-15959784 on Build GRCh38
rs34220997 Chr.12: 15959842-15959842 on Build GRCh38
rs772473455 Chr.12: 15959878-15959878 on Build GRCh38

View Full Product Details