Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68022826-68023093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX28
Entrez Gene ID
55794
Gene Name
DEAD-box helicase 28
Gene Aliases
MDDX28
UniGene
Hs.458313
Gene Chromosome Location
Chr.16: 68021274-68023867 on Build GRCh38
Transcripts
NM_018380.3 NM_001271762.1 NM_001271763.1 NM_017803.4

Gene Symbol
DUS2
Entrez Gene ID
54920
Gene Name
dihydrouridine synthase 2
Gene Aliases
DUS2L, SMM1, URLC8
UniGene
Hs.534460
Gene Chromosome Location
Chr.16: 68022944-68079323 on Build GRCh38
Transcripts
NM_018380.3 NM_001271762.1 NM_001271763.1 NM_017803.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6363099 c.227T>G p.L76W Chr.16: 68022976-68022976 on Build GRCh38
3511172 c.144G>A p.Q48Q Chr.16: 68023059-68023059 on Build GRCh38
6641695 c.124C>A p.L42I Chr.16: 68023079-68023079 on Build GRCh38
4638958 c.240G>A p.A80A Chr.16: 68022963-68022963 on Build GRCh38
703798 c.157C>T p.R53W Chr.16: 68023046-68023046 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971328072 Chr.16: 68022989-68022989 on Build GRCh38
rs747604269 Chr.16: 68023037-68023037 on Build GRCh38
rs764317601 Chr.16: 68023071-68023071 on Build GRCh38
rs765330276 Chr.16: 68023061-68023061 on Build GRCh38
rs750716775 Chr.16: 68022882-68022882 on Build GRCh38
rs201701226 Chr.16: 68022954-68022954 on Build GRCh38
rs189530871 Chr.16: 68022861-68022861 on Build GRCh38
rs940845656 Chr.16: 68022892-68022892 on Build GRCh38
rs902120683 Chr.16: 68022918-68022918 on Build GRCh38
rs748964960 Chr.16: 68023007-68023007 on Build GRCh38
rs369855270 Chr.16: 68022953-68022953 on Build GRCh38
rs1015693133 Chr.16: 68022979-68022979 on Build GRCh38
rs778920793 Chr.16: 68022896-68022896 on Build GRCh38
rs375274355 Chr.16: 68022977-68022978 on Build GRCh38
rs928715177 Chr.16: 68023022-68023022 on Build GRCh38
rs749048192 Chr.16: 68022865-68022866 on Build GRCh38
rs201107769 Chr.16: 68022985-68022985 on Build GRCh38
rs774064363 Chr.16: 68023030-68023030 on Build GRCh38
rs752083060 Chr.16: 68022870-68022870 on Build GRCh38
rs1048766236 Chr.16: 68022873-68022873 on Build GRCh38
rs200300816 Chr.16: 68022994-68022994 on Build GRCh38
rs775028529 Chr.16: 68022858-68022858 on Build GRCh38
rs776637544 Chr.16: 68022848-68022848 on Build GRCh38
rs776796410 Chr.16: 68023057-68023057 on Build GRCh38
rs766763886 Chr.16: 68023056-68023056 on Build GRCh38
rs768123979 Chr.16: 68023009-68023009 on Build GRCh38
rs755067862 Chr.16: 68022890-68022890 on Build GRCh38
rs553471941 Chr.16: 68022948-68022948 on Build GRCh38
rs754080255 Chr.16: 68022969-68022969 on Build GRCh38
rs202069030 Chr.16: 68023050-68023050 on Build GRCh38
rs112237019 Chr.16: 68022893-68022893 on Build GRCh38
rs781293191 Chr.16: 68022908-68022908 on Build GRCh38
rs755226130 Chr.16: 68022970-68022970 on Build GRCh38
rs557840864 Chr.16: 68022981-68022981 on Build GRCh38
rs371263024 Chr.16: 68023020-68023020 on Build GRCh38
rs956082572 Chr.16: 68022956-68022956 on Build GRCh38
rs780368968 Chr.16: 68023033-68023033 on Build GRCh38
rs765502635 Chr.16: 68022972-68022972 on Build GRCh38
rs762363062 Chr.16: 68022871-68022871 on Build GRCh38
rs754051880 Chr.16: 68022888-68022888 on Build GRCh38
rs773726597 Chr.16: 68022950-68022950 on Build GRCh38
rs780197191 Chr.16: 68022887-68022887 on Build GRCh38
rs770839264 Chr.16: 68022847-68022847 on Build GRCh38
rs768197148 Chr.16: 68022927-68022927 on Build GRCh38
rs745840315 Chr.16: 68022988-68022988 on Build GRCh38
rs745609429 Chr.16: 68022912-68022912 on Build GRCh38
rs771598308 Chr.16: 68023048-68023048 on Build GRCh38
rs759959392 Chr.16: 68023059-68023059 on Build GRCh38
rs549305821 Chr.16: 68022886-68022886 on Build GRCh38
rs184276673 Chr.16: 68022929-68022929 on Build GRCh38
rs770927214 Chr.16: 68022905-68022905 on Build GRCh38
rs752999743 Chr.16: 68023064-68023064 on Build GRCh38
rs769514331 Chr.16: 68022853-68022853 on Build GRCh38
rs779768845 Chr.16: 68023006-68023006 on Build GRCh38
rs758521504 Chr.16: 68023067-68023067 on Build GRCh38
rs767860967 Chr.16: 68022879-68022879 on Build GRCh38
rs535461824 Chr.16: 68022947-68022947 on Build GRCh38
rs763551974 Chr.16: 68022869-68022869 on Build GRCh38
rs200648087 Chr.16: 68022926-68022926 on Build GRCh38
rs758720620 Chr.16: 68022994-68022995 on Build GRCh38
rs759885538 Chr.16: 68022920-68022920 on Build GRCh38
rs752768990 Chr.16: 68022980-68022980 on Build GRCh38
rs746964562 Chr.16: 68022897-68022897 on Build GRCh38
rs759411290 Chr.16: 68022849-68022849 on Build GRCh38

View Full Product Details