Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 47078816-47079075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMIGO2
Entrez Gene ID
347902
Gene Name
adhesion molecule with Ig like domain 2
Gene Aliases
ALI1, AMIGO-2, DEGA
UniGene
Hs.121520
Gene Chromosome Location
Chr.12: 47075707-47080826 on Build GRCh38
Transcripts
NM_001143668.1 NM_181847.4 XM_005268837.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5841015 c.110C>T p.A37V Chr.12: 47078893-47078893 on Build GRCh38
5027579 c.107G>A p.G36D Chr.12: 47078896-47078896 on Build GRCh38
7080047 c.38G>A p.G13E Chr.12: 47078965-47078965 on Build GRCh38
5929364 c.64G>A p.E22K Chr.12: 47078939-47078939 on Build GRCh38
7113164 c.93G>A p.V31V Chr.12: 47078910-47078910 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757997435 Chr.12: 47079042-47079042 on Build GRCh38
rs760568048 Chr.12: 47079007-47079007 on Build GRCh38
rs776343317 Chr.12: 47079014-47079014 on Build GRCh38
rs769967370 Chr.12: 47078987-47078987 on Build GRCh38
rs527422309 Chr.12: 47078995-47078995 on Build GRCh38
rs368728537 Chr.12: 47078866-47078866 on Build GRCh38
rs556169324 Chr.12: 47078912-47078912 on Build GRCh38
rs755894400 Chr.12: 47078852-47078852 on Build GRCh38
rs770025296 Chr.12: 47078911-47078911 on Build GRCh38
rs772759106 Chr.12: 47078880-47078880 on Build GRCh38
rs753370490 Chr.12: 47078859-47078859 on Build GRCh38
rs78132511 Chr.12: 47079027-47079027 on Build GRCh38
rs759366874 Chr.12: 47079017-47079017 on Build GRCh38
rs753562011 Chr.12: 47078957-47078957 on Build GRCh38
rs768662432 Chr.12: 47078998-47078998 on Build GRCh38
rs200841027 Chr.12: 47079030-47079030 on Build GRCh38
rs761120703 Chr.12: 47079029-47079030 on Build GRCh38
rs35667451 Chr.12: 47078910-47078910 on Build GRCh38
rs557234455 Chr.12: 47078838-47078838 on Build GRCh38
rs778429412 Chr.12: 47078978-47078978 on Build GRCh38
rs1037175131 Chr.12: 47078929-47078929 on Build GRCh38
rs763650753 Chr.12: 47079043-47079043 on Build GRCh38
rs760473930 Chr.12: 47078953-47078953 on Build GRCh38
rs938044641 Chr.12: 47078906-47078906 on Build GRCh38
rs781449690 Chr.12: 47078982-47078982 on Build GRCh38
rs1049497177 Chr.12: 47078997-47078997 on Build GRCh38
rs372752192 Chr.12: 47079010-47079010 on Build GRCh38
rs773063621 Chr.12: 47078948-47078948 on Build GRCh38
rs746220242 Chr.12: 47078986-47078986 on Build GRCh38
rs776926522 Chr.12: 47078901-47078901 on Build GRCh38
rs573867386 Chr.12: 47078881-47078881 on Build GRCh38
rs35963513 Chr.12: 47078952-47078952 on Build GRCh38
rs764925535 Chr.12: 47078958-47078958 on Build GRCh38
rs373107438 Chr.12: 47079045-47079045 on Build GRCh38
rs79830988 Chr.12: 47079026-47079026 on Build GRCh38
rs779759742 Chr.12: 47078854-47078854 on Build GRCh38
rs201981890 Chr.12: 47078918-47078918 on Build GRCh38
rs147185355 Chr.12: 47078969-47078969 on Build GRCh38
rs757979001 Chr.12: 47078971-47078971 on Build GRCh38
rs775772687 Chr.12: 47078988-47078988 on Build GRCh38
rs747667219 Chr.12: 47078874-47078874 on Build GRCh38
rs757260518 Chr.12: 47078983-47078983 on Build GRCh38
rs369336872 Chr.12: 47078939-47078939 on Build GRCh38
rs778412775 Chr.12: 47078885-47078885 on Build GRCh38
rs113339201 Chr.12: 47078981-47078981 on Build GRCh38
rs747462184 Chr.12: 47078979-47078979 on Build GRCh38
rs139517038 Chr.12: 47079004-47079004 on Build GRCh38
rs760892641 Chr.12: 47078883-47078883 on Build GRCh38
rs944227759 Chr.12: 47078996-47078998 on Build GRCh38
rs766036941 Chr.12: 47078955-47078955 on Build GRCh38
rs372852092 Chr.12: 47078886-47078886 on Build GRCh38
rs111900491 Chr.12: 47078944-47078944 on Build GRCh38
rs771356711 Chr.12: 47078900-47078900 on Build GRCh38
rs749370909 Chr.12: 47078992-47078992 on Build GRCh38

View Full Product Details