Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80996990-80997217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP6
Entrez Gene ID
79643
Gene Name
charged multivesicular body protein 6
Gene Aliases
VPS20
UniGene
Hs.514560
Gene Chromosome Location
Chr.17: 80991841-81000133 on Build GRCh38
Transcripts
NM_024591.4 XM_005257668.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4856905 c.364G>C p.E122Q Chr.17: 80997022-80997022 on Build GRCh38
985768 c.389C>T p.T130M Chr.17: 80997047-80997047 on Build GRCh38
985767 c.384C>T p.D128D Chr.17: 80997042-80997042 on Build GRCh38
3712475 c.387G>A p.E129E Chr.17: 80997045-80997045 on Build GRCh38
3523463 c.364G>A p.E122K Chr.17: 80997022-80997022 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369414852 Chr.17: 80997042-80997042 on Build GRCh38
rs769508597 Chr.17: 80997058-80997058 on Build GRCh38
rs1005653693 Chr.17: 80997189-80997189 on Build GRCh38
rs537210333 Chr.17: 80997148-80997148 on Build GRCh38
rs376878102 Chr.17: 80997081-80997081 on Build GRCh38
rs899472991 Chr.17: 80997191-80997191 on Build GRCh38
rs1016118374 Chr.17: 80997197-80997197 on Build GRCh38
rs552485187 Chr.17: 80997163-80997163 on Build GRCh38
rs569856671 Chr.17: 80997130-80997130 on Build GRCh38
rs764679129 Chr.17: 80997017-80997017 on Build GRCh38
rs552536018 Chr.17: 80997185-80997185 on Build GRCh38
rs777212296 Chr.17: 80997090-80997090 on Build GRCh38
rs867192457 Chr.17: 80997073-80997073 on Build GRCh38
rs571705472 Chr.17: 80997071-80997071 on Build GRCh38
rs750750575 Chr.17: 80997118-80997118 on Build GRCh38
rs979496082 Chr.17: 80997040-80997040 on Build GRCh38
rs200778781 Chr.17: 80997015-80997015 on Build GRCh38
rs748247959 Chr.17: 80997122-80997124 on Build GRCh38
rs757660097 Chr.17: 80997032-80997032 on Build GRCh38
rs1046676108 Chr.17: 80997145-80997145 on Build GRCh38
rs761287804 Chr.17: 80997016-80997016 on Build GRCh38
rs772807455 Chr.17: 80997008-80997008 on Build GRCh38
rs1002335598 Chr.17: 80997162-80997162 on Build GRCh38
rs4969334 Chr.17: 80997109-80997109 on Build GRCh38
rs920097501 Chr.17: 80997129-80997129 on Build GRCh38
rs891476875 Chr.17: 80997125-80997125 on Build GRCh38
rs931459276 Chr.17: 80997141-80997141 on Build GRCh38
rs148035263 Chr.17: 80997043-80997043 on Build GRCh38
rs777413976 Chr.17: 80997068-80997068 on Build GRCh38
rs765854602 Chr.17: 80997014-80997014 on Build GRCh38
rs762369762 Chr.17: 80997010-80997010 on Build GRCh38
rs150171612 Chr.17: 80997057-80997057 on Build GRCh38
rs754281461 Chr.17: 80997022-80997022 on Build GRCh38
rs374165984 Chr.17: 80997070-80997070 on Build GRCh38
rs769322225 Chr.17: 80997089-80997089 on Build GRCh38
rs750603328 Chr.17: 80997035-80997035 on Build GRCh38
rs534627562 Chr.17: 80997180-80997180 on Build GRCh38
rs371590901 Chr.17: 80997083-80997083 on Build GRCh38
rs114525609 Chr.17: 80997091-80997091 on Build GRCh38
rs906824677 Chr.17: 80997158-80997158 on Build GRCh38
rs377068389 Chr.17: 80997033-80997033 on Build GRCh38
rs1046885388 Chr.17: 80997151-80997151 on Build GRCh38
rs555274393 Chr.17: 80997159-80997159 on Build GRCh38
rs140815434 Chr.17: 80997048-80997048 on Build GRCh38
rs751560399 Chr.17: 80997047-80997047 on Build GRCh38

View Full Product Details