Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 34246799-34247052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf1
Entrez Gene ID
221491
Gene Name
chromosome 6 open reading frame 1
Gene Aliases
LBH
UniGene
Hs.381300
Gene Chromosome Location
Chr.6: 34246380-34249108 on Build GRCh38
Transcripts
NM_001008703.2 NM_001008704.2 NM_001287396.1 NM_001287397.1 NM_001287398.1 NM_178508.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3873357 c.122C>T p.P41L Chr.6: 34247047-34247047 on Build GRCh38
6208084 c.131A>C p.Y44S Chr.6: 34246863-34246863 on Build GRCh38
1294325 c.179T>C p.M60T Chr.6: 34246815-34246815 on Build GRCh38
1329738 c.181T>G p.S61A Chr.6: 34246813-34246813 on Build GRCh38
6535904 c.143G>C p.R48T Chr.6: 34246851-34246851 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752471956 Chr.6: 34246863-34246864 on Build GRCh38
rs777675776 Chr.6: 34246860-34246860 on Build GRCh38
rs367811394 Chr.6: 34246858-34246858 on Build GRCh38
rs773683356 Chr.6: 34247003-34247003 on Build GRCh38
rs3216050 Chr.6: 34246933-34246934 on Build GRCh38
rs956364893 Chr.6: 34246907-34246907 on Build GRCh38
rs752612863 Chr.6: 34247031-34247031 on Build GRCh38
rs764233280 Chr.6: 34246909-34246909 on Build GRCh38
rs765132214 Chr.6: 34246832-34246837 on Build GRCh38
rs976795670 Chr.6: 34246992-34246992 on Build GRCh38
rs746067941 Chr.6: 34246863-34246863 on Build GRCh38
rs967166678 Chr.6: 34246987-34246987 on Build GRCh38
rs748703186 Chr.6: 34246906-34246906 on Build GRCh38
rs766070074 Chr.6: 34247011-34247011 on Build GRCh38
rs762802801 Chr.6: 34246876-34246878 on Build GRCh38
rs386700055 Chr.6: 34246958-34246960 on Build GRCh38
rs969359793 Chr.6: 34246942-34246942 on Build GRCh38
rs765182842 Chr.6: 34246854-34246854 on Build GRCh38
rs752139148 Chr.6: 34246873-34246873 on Build GRCh38
rs764770220 Chr.6: 34247030-34247030 on Build GRCh38
rs776072462 Chr.6: 34247024-34247024 on Build GRCh38
rs777810489 Chr.6: 34246875-34246875 on Build GRCh38
rs397840178 Chr.6: 34246934-34246935 on Build GRCh38
rs920992679 Chr.6: 34246857-34246857 on Build GRCh38
rs559657962 Chr.6: 34246977-34246977 on Build GRCh38
rs750012431 Chr.6: 34246833-34246833 on Build GRCh38
rs765802250 Chr.6: 34246842-34246842 on Build GRCh38
rs754852922 Chr.6: 34246848-34246848 on Build GRCh38
rs370960321 Chr.6: 34247005-34247005 on Build GRCh38
rs749388885 Chr.6: 34246902-34246902 on Build GRCh38
rs4711389 Chr.6: 34246893-34246893 on Build GRCh38
rs1033793365 Chr.6: 34246943-34246943 on Build GRCh38
rs548595794 Chr.6: 34247013-34247013 on Build GRCh38
rs375245462 Chr.6: 34246832-34246832 on Build GRCh38
rs772301810 Chr.6: 34246917-34246917 on Build GRCh38
rs935620773 Chr.6: 34246839-34246839 on Build GRCh38
rs1002157975 Chr.6: 34246927-34246927 on Build GRCh38
rs371338846 Chr.6: 34246868-34246868 on Build GRCh38
rs769234473 Chr.6: 34246903-34246903 on Build GRCh38
rs530434268 Chr.6: 34246985-34246985 on Build GRCh38
rs756420796 Chr.6: 34246882-34246882 on Build GRCh38
rs201938239 Chr.6: 34246831-34246831 on Build GRCh38
rs989543049 Chr.6: 34246926-34246926 on Build GRCh38
rs753393548 Chr.6: 34246843-34246843 on Build GRCh38
rs925613731 Chr.6: 34246836-34246836 on Build GRCh38
rs760425041 Chr.6: 34246840-34246840 on Build GRCh38
rs571773263 Chr.6: 34246948-34246948 on Build GRCh38
rs759079915 Chr.6: 34247025-34247025 on Build GRCh38
rs887258731 Chr.6: 34246876-34246876 on Build GRCh38
rs548034843 Chr.6: 34246960-34246960 on Build GRCh38
rs376787037 Chr.6: 34246891-34246891 on Build GRCh38
rs114233485 Chr.6: 34246958-34246958 on Build GRCh38
rs763796441 Chr.6: 34247032-34247032 on Build GRCh38
rs775027280 Chr.6: 34246904-34246904 on Build GRCh38

View Full Product Details