Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 229518128-229518387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB10
Entrez Gene ID
23456
Gene Name
ATP binding cassette subfamily B member 10
Gene Aliases
EST20237, M-ABC2, MTABC2
UniGene
Hs.17614
Gene Chromosome Location
Chr.1: 229516582-229558987 on Build GRCh38
Transcripts
NM_012089.2 XM_011544135.2 XM_011544136.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
905263 c.2151G>T p.E717D Chr.1: 229518245-229518245 on Build GRCh38
6541941 c.2048G>A p.R683K Chr.1: 229518348-229518348 on Build GRCh38
1134837 c.2142A>G p.K714K Chr.1: 229518254-229518254 on Build GRCh38
4400020 c.2060T>G p.V687G Chr.1: 229518336-229518336 on Build GRCh38
6326077 c.2017C>T p.Q673* Chr.1: 229518379-229518379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970122557 Chr.1: 229518357-229518358 on Build GRCh38
rs911596560 Chr.1: 229518244-229518244 on Build GRCh38
rs771365184 Chr.1: 229518341-229518341 on Build GRCh38
rs756498385 Chr.1: 229518323-229518323 on Build GRCh38
rs768931908 Chr.1: 229518167-229518167 on Build GRCh38
rs201773661 Chr.1: 229518351-229518351 on Build GRCh38
rs377159208 Chr.1: 229518253-229518253 on Build GRCh38
rs748496881 Chr.1: 229518184-229518184 on Build GRCh38
rs546462477 Chr.1: 229518292-229518292 on Build GRCh38
rs145356391 Chr.1: 229518223-229518223 on Build GRCh38
rs750370909 Chr.1: 229518268-229518268 on Build GRCh38
rs781300607 Chr.1: 229518182-229518182 on Build GRCh38
rs747098205 Chr.1: 229518158-229518158 on Build GRCh38
rs770623136 Chr.1: 229518354-229518354 on Build GRCh38
rs767831138 Chr.1: 229518187-229518187 on Build GRCh38
rs963573449 Chr.1: 229518174-229518174 on Build GRCh38
rs144991345 Chr.1: 229518261-229518261 on Build GRCh38
rs758351353 Chr.1: 229518279-229518279 on Build GRCh38
rs766423760 Chr.1: 229518290-229518290 on Build GRCh38
rs141003292 Chr.1: 229518224-229518224 on Build GRCh38
rs770106506 Chr.1: 229518202-229518202 on Build GRCh38
rs185229946 Chr.1: 229518363-229518363 on Build GRCh38
rs746790498 Chr.1: 229518311-229518311 on Build GRCh38
rs778060314 Chr.1: 229518329-229518329 on Build GRCh38
rs572050882 Chr.1: 229518314-229518314 on Build GRCh38
rs370644459 Chr.1: 229518264-229518264 on Build GRCh38
rs760185728 Chr.1: 229518228-229518228 on Build GRCh38
rs773740175 Chr.1: 229518211-229518211 on Build GRCh38
rs117077784 Chr.1: 229518345-229518345 on Build GRCh38
rs763766243 Chr.1: 229518248-229518248 on Build GRCh38
rs781455022 Chr.1: 229518313-229518313 on Build GRCh38
rs749669414 Chr.1: 229518334-229518334 on Build GRCh38
rs115488729 Chr.1: 229518306-229518306 on Build GRCh38

View Full Product Details