Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1455158-1455415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APC2
Entrez Gene ID
10297
Gene Name
APC2, WNT signaling pathway regulator
Gene Aliases
APCL
UniGene
Hs.446376
Gene Chromosome Location
Chr.19: 1446268-1473244 on Build GRCh38
Transcripts
NM_005883.2 XM_005259475.2 XM_006722607.2 XM_006722608.3 XM_006722609.3 XM_006722610.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5086443 c.440_442delAGG p.E149delE Chr.19: 1455175-1455177 on Build GRCh38
7355939 c.468C>G p.Y156* Chr.19: 1455203-1455203 on Build GRCh38
6903356 c.497delT p.L166fs*17 Chr.19: 1455232-1455232 on Build GRCh38
6376613 c.485T>C p.L162P Chr.19: 1455220-1455220 on Build GRCh38
5749812 c.446_448delAGA p.K150delK Chr.19: 1455181-1455183 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370125088 Chr.19: 1455368-1455368 on Build GRCh38
rs1024880349 Chr.19: 1455236-1455236 on Build GRCh38
rs528448967 Chr.19: 1455375-1455375 on Build GRCh38
rs971539422 Chr.19: 1455248-1455248 on Build GRCh38
rs752311235 Chr.19: 1455247-1455247 on Build GRCh38
rs754123761 Chr.19: 1455203-1455203 on Build GRCh38
rs1037275589 Chr.19: 1455343-1455343 on Build GRCh38
rs576724127 Chr.19: 1455267-1455267 on Build GRCh38
rs768840660 Chr.19: 1455292-1455292 on Build GRCh38
rs746240851 Chr.19: 1455309-1455309 on Build GRCh38
rs780790406 Chr.19: 1455232-1455232 on Build GRCh38
rs745764289 Chr.19: 1455282-1455282 on Build GRCh38
rs372827362 Chr.19: 1455336-1455336 on Build GRCh38
rs780018629 Chr.19: 1455387-1455387 on Build GRCh38
rs767057669 Chr.19: 1455340-1455340 on Build GRCh38
rs765538031 Chr.19: 1455219-1455219 on Build GRCh38
rs894663477 Chr.19: 1455217-1455217 on Build GRCh38
rs933805250 Chr.19: 1455386-1455386 on Build GRCh38
rs775545468 Chr.19: 1455286-1455286 on Build GRCh38
rs562328926 Chr.19: 1455318-1455318 on Build GRCh38
rs377021589 Chr.19: 1455360-1455360 on Build GRCh38
rs761968822 Chr.19: 1455304-1455304 on Build GRCh38
rs773461825 Chr.19: 1455333-1455333 on Build GRCh38
rs778759824 Chr.19: 1455366-1455366 on Build GRCh38
rs757498007 Chr.19: 1455184-1455186 on Build GRCh38
rs764369973 Chr.19: 1455200-1455200 on Build GRCh38
rs911708482 Chr.19: 1455397-1455397 on Build GRCh38
rs746968102 Chr.19: 1455389-1455389 on Build GRCh38
rs140079855 Chr.19: 1455218-1455218 on Build GRCh38
rs370854425 Chr.19: 1455265-1455265 on Build GRCh38
rs776130430 Chr.19: 1455368-1455369 on Build GRCh38
rs558389908 Chr.19: 1455244-1455244 on Build GRCh38
rs781465656 Chr.19: 1455302-1455303 on Build GRCh38
rs372277299 Chr.19: 1455321-1455321 on Build GRCh38
rs756874379 Chr.19: 1455354-1455354 on Build GRCh38
rs747135563 Chr.19: 1455287-1455287 on Build GRCh38
rs763545966 Chr.19: 1455348-1455348 on Build GRCh38
rs758977342 Chr.19: 1455226-1455226 on Build GRCh38
rs760878340 Chr.19: 1455191-1455191 on Build GRCh38
rs778577691 Chr.19: 1455279-1455279 on Build GRCh38
rs561230877 Chr.19: 1455350-1455350 on Build GRCh38
rs768678791 Chr.19: 1455391-1455391 on Build GRCh38
rs757037018 Chr.19: 1455275-1455275 on Build GRCh38
rs752221383 Chr.19: 1455344-1455344 on Build GRCh38
rs748917200 Chr.19: 1455272-1455272 on Build GRCh38
rs770271842 Chr.19: 1455362-1455362 on Build GRCh38
rs915652338 Chr.19: 1455222-1455222 on Build GRCh38
rs544127737 Chr.19: 1455299-1455299 on Build GRCh38
rs948417851 Chr.19: 1455229-1455229 on Build GRCh38
rs758303503 Chr.19: 1455371-1455371 on Build GRCh38
rs77484699 Chr.19: 1455319-1455319 on Build GRCh38
rs760020769 Chr.19: 1455246-1455246 on Build GRCh38
rs755637764 Chr.19: 1455347-1455347 on Build GRCh38
rs765567854 Chr.19: 1455309-1455309 on Build GRCh38
rs750899180 Chr.19: 1455224-1455224 on Build GRCh38

View Full Product Details