Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 92500239-92500512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf32
Entrez Gene ID
145858
Gene Name
chromosome 15 open reading frame 32
Gene Aliases
-
UniGene
Hs.367879
Gene Chromosome Location
Chr.15: 92465762-92501117 on Build GRCh38
Transcripts
NM_001301106.1 NM_153040.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1375529 c.527G>T p.C176F Chr.15: 92500390-92500390 on Build GRCh38
5942617 c.495-5T>A p.? Chr.15: 92500353-92500353 on Build GRCh38
3505515 c.496G>A p.E166K Chr.15: 92500359-92500359 on Build GRCh38
4595398 c.497A>G p.E166G Chr.15: 92500360-92500360 on Build GRCh38
344385 c.499C>A p.H167N Chr.15: 92500362-92500362 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs969271100 Chr.15: 92500465-92500465 on Build GRCh38
rs556183528 Chr.15: 92500459-92500459 on Build GRCh38
rs370529720 Chr.15: 92500289-92500289 on Build GRCh38
rs775058816 Chr.15: 92500350-92500350 on Build GRCh38
rs761479676 Chr.15: 92500369-92500369 on Build GRCh38
rs765306269 Chr.15: 92500325-92500325 on Build GRCh38
rs924909522 Chr.15: 92500288-92500288 on Build GRCh38
rs752810746 Chr.15: 92500426-92500426 on Build GRCh38
rs1023448318 Chr.15: 92500407-92500407 on Build GRCh38
rs762601691 Chr.15: 92500311-92500311 on Build GRCh38
rs375715557 Chr.15: 92500345-92500345 on Build GRCh38
rs977592634 Chr.15: 92500480-92500480 on Build GRCh38
rs774028676 Chr.15: 92500364-92500364 on Build GRCh38
rs769049564 Chr.15: 92500405-92500405 on Build GRCh38
rs369668372 Chr.15: 92500400-92500400 on Build GRCh38
rs113823034 Chr.15: 92500406-92500406 on Build GRCh38
rs763805503 Chr.15: 92500312-92500312 on Build GRCh38
rs766296548 Chr.15: 92500415-92500415 on Build GRCh38
rs751782328 Chr.15: 92500437-92500437 on Build GRCh38
rs116503901 Chr.15: 92500411-92500411 on Build GRCh38
rs533248348 Chr.15: 92500283-92500283 on Build GRCh38
rs372046606 Chr.15: 92500307-92500307 on Build GRCh38
rs956486731 Chr.15: 92500306-92500306 on Build GRCh38
rs555385111 Chr.15: 92500323-92500323 on Build GRCh38
rs201939844 Chr.15: 92500332-92500332 on Build GRCh38
rs750338738 Chr.15: 92500322-92500322 on Build GRCh38
rs767297455 Chr.15: 92500372-92500372 on Build GRCh38
rs748609097 Chr.15: 92500478-92500478 on Build GRCh38
rs768414814 Chr.15: 92500340-92500340 on Build GRCh38
rs781431643 Chr.15: 92500341-92500341 on Build GRCh38
rs748851364 Chr.15: 92500357-92500357 on Build GRCh38
rs7178722 Chr.15: 92500315-92500315 on Build GRCh38
rs912734751 Chr.15: 92500314-92500314 on Build GRCh38
rs953418021 Chr.15: 92500448-92500448 on Build GRCh38
rs543669748 Chr.15: 92500452-92500452 on Build GRCh38
rs764380887 Chr.15: 92500432-92500432 on Build GRCh38
rs1007590070 Chr.15: 92500450-92500450 on Build GRCh38
rs758601786 Chr.15: 92500431-92500431 on Build GRCh38
rs781781067 Chr.15: 92500453-92500453 on Build GRCh38
rs147870364 Chr.15: 92500334-92500334 on Build GRCh38
rs146402235 Chr.15: 92500317-92500317 on Build GRCh38
rs768271793 Chr.15: 92500359-92500359 on Build GRCh38
rs371292270 Chr.15: 92500324-92500324 on Build GRCh38
rs969098210 Chr.15: 92500280-92500280 on Build GRCh38
rs34262682 Chr.15: 92500296-92500297 on Build GRCh38
rs767406751 Chr.15: 92500320-92500320 on Build GRCh38
rs182909589 Chr.15: 92500331-92500331 on Build GRCh38
rs780189166 Chr.15: 92500423-92500428 on Build GRCh38
rs746933155 Chr.15: 92500431-92500437 on Build GRCh38
rs34102544 Chr.15: 92500310-92500311 on Build GRCh38
rs746328071 Chr.15: 92500342-92500342 on Build GRCh38
rs376196928 Chr.15: 92500338-92500338 on Build GRCh38
rs757736082 Chr.15: 92500339-92500339 on Build GRCh38
rs772899841 Chr.15: 92500404-92500404 on Build GRCh38

View Full Product Details