Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 29030532-29030803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPVL
Entrez Gene ID
54504
Gene Name
carboxypeptidase, vitellogenic like
Gene Aliases
HVLP
UniGene
Hs.233389
Gene Chromosome Location
Chr.7: 28995630-29146537 on Build GRCh38
Transcripts
NM_019029.2 NM_031311.3 XM_011515437.1 XM_017012366.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3027684 c.1240G>A p.A414T Chr.7: 29030657-29030657 on Build GRCh38
6177713 c.1305G>T p.A435A Chr.7: 29030592-29030592 on Build GRCh38
3027686 c.1199T>A p.L400* Chr.7: 29030698-29030698 on Build GRCh38
5872559 c.1285G>A p.A429T Chr.7: 29030612-29030612 on Build GRCh38
389939 c.1169T>C p.I390T Chr.7: 29030728-29030728 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759176203 Chr.7: 29030565-29030565 on Build GRCh38
rs766374508 Chr.7: 29030717-29030717 on Build GRCh38
rs780931131 Chr.7: 29030592-29030592 on Build GRCh38
rs774789923 Chr.7: 29030590-29030590 on Build GRCh38
rs751478780 Chr.7: 29030746-29030746 on Build GRCh38
rs7313 Chr.7: 29030593-29030593 on Build GRCh38
rs780727520 Chr.7: 29030571-29030572 on Build GRCh38
rs773880380 Chr.7: 29030705-29030705 on Build GRCh38
rs768929266 Chr.7: 29030577-29030577 on Build GRCh38
rs945652017 Chr.7: 29030710-29030710 on Build GRCh38
rs758188906 Chr.7: 29030634-29030634 on Build GRCh38
rs561008600 Chr.7: 29030680-29030680 on Build GRCh38
rs776192680 Chr.7: 29030564-29030564 on Build GRCh38
rs758236007 Chr.7: 29030735-29030735 on Build GRCh38
rs759525082 Chr.7: 29030727-29030727 on Build GRCh38
rs1052200 Chr.7: 29030704-29030704 on Build GRCh38
rs763825813 Chr.7: 29030740-29030740 on Build GRCh38
rs752888902 Chr.7: 29030733-29030733 on Build GRCh38
rs750959204 Chr.7: 29030594-29030594 on Build GRCh38
rs760718602 Chr.7: 29030599-29030599 on Build GRCh38
rs1042506922 Chr.7: 29030664-29030664 on Build GRCh38
rs375729583 Chr.7: 29030749-29030749 on Build GRCh38
rs371059469 Chr.7: 29030600-29030600 on Build GRCh38
rs35699738 Chr.7: 29030751-29030751 on Build GRCh38
rs372394394 Chr.7: 29030699-29030699 on Build GRCh38
rs775268596 Chr.7: 29030695-29030695 on Build GRCh38
rs146520943 Chr.7: 29030762-29030762 on Build GRCh38
rs746717432 Chr.7: 29030769-29030769 on Build GRCh38
rs779892091 Chr.7: 29030777-29030777 on Build GRCh38
rs772597879 Chr.7: 29030780-29030780 on Build GRCh38
rs778697580 Chr.7: 29030608-29030608 on Build GRCh38
rs765512457 Chr.7: 29030730-29030730 on Build GRCh38
rs752549832 Chr.7: 29030615-29030615 on Build GRCh38
rs776719397 Chr.7: 29030726-29030726 on Build GRCh38
rs755161934 Chr.7: 29030607-29030607 on Build GRCh38
rs112539958 Chr.7: 29030778-29030778 on Build GRCh38
rs770366520 Chr.7: 29030671-29030671 on Build GRCh38
rs376419169 Chr.7: 29030551-29030551 on Build GRCh38
rs769429493 Chr.7: 29030691-29030691 on Build GRCh38
rs745441635 Chr.7: 29030752-29030752 on Build GRCh38
rs746941326 Chr.7: 29030640-29030640 on Build GRCh38
rs761204292 Chr.7: 29030707-29030707 on Build GRCh38
rs949471035 Chr.7: 29030651-29030651 on Build GRCh38
rs745508081 Chr.7: 29030684-29030684 on Build GRCh38
rs770440563 Chr.7: 29030556-29030556 on Build GRCh38
rs777730030 Chr.7: 29030636-29030636 on Build GRCh38

View Full Product Details