Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44287025-44287294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIRE
Entrez Gene ID
326
Gene Name
autoimmune regulator
Gene Aliases
AIRE1, APECED, APS1, APSI, PGA1
UniGene
Hs.129829
Gene Chromosome Location
Chr.21: 44285838-44298219 on Build GRCh38
Transcripts
NM_000383.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6597294 c.413C>A p.A138D Chr.21: 44287083-44287083 on Build GRCh38
2820599 c.371C>T p.P124L Chr.21: 44287041-44287041 on Build GRCh38
419672 c.442C>G p.P148A Chr.21: 44287112-44287112 on Build GRCh38
6597293 c.372G>A p.P124P Chr.21: 44287042-44287042 on Build GRCh38
6597289 c.423C>T p.A141A Chr.21: 44287093-44287093 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569224551 Chr.21: 44287138-44287138 on Build GRCh38
rs1012724825 Chr.21: 44287158-44287158 on Build GRCh38
rs776770003 Chr.21: 44287263-44287264 on Build GRCh38
rs753974982 Chr.21: 44287166-44287166 on Build GRCh38
rs763642554 Chr.21: 44287137-44287137 on Build GRCh38
rs786204478 Chr.21: 44287135-44287135 on Build GRCh38
rs771314896 Chr.21: 44287124-44287124 on Build GRCh38
rs941659310 Chr.21: 44287238-44287238 on Build GRCh38
rs545287094 Chr.21: 44287231-44287231 on Build GRCh38
rs775117249 Chr.21: 44287042-44287042 on Build GRCh38
rs754211503 Chr.21: 44287248-44287248 on Build GRCh38
rs879189408 Chr.21: 44287230-44287230 on Build GRCh38
rs763835269 Chr.21: 44287064-44287064 on Build GRCh38
rs937627063 Chr.21: 44287214-44287214 on Build GRCh38
rs932783037 Chr.21: 44287068-44287068 on Build GRCh38
rs577544633 Chr.21: 44287227-44287227 on Build GRCh38
rs201965760 Chr.21: 44287086-44287086 on Build GRCh38
rs572002344 Chr.21: 44287093-44287093 on Build GRCh38
rs121434256 Chr.21: 44287085-44287085 on Build GRCh38
rs761329134 Chr.21: 44287149-44287149 on Build GRCh38
rs374704178 Chr.21: 44287117-44287117 on Build GRCh38
rs755062200 Chr.21: 44287168-44287168 on Build GRCh38
rs778926390 Chr.21: 44287169-44287169 on Build GRCh38
rs912713336 Chr.21: 44287256-44287256 on Build GRCh38
rs779092108 Chr.21: 44287095-44287095 on Build GRCh38
rs552736654 Chr.21: 44287213-44287213 on Build GRCh38
rs997396625 Chr.21: 44287129-44287129 on Build GRCh38
rs1045928758 Chr.21: 44287207-44287207 on Build GRCh38
rs758527824 Chr.21: 44287173-44287173 on Build GRCh38
rs536357179 Chr.21: 44287170-44287170 on Build GRCh38
rs567147078 Chr.21: 44287179-44287179 on Build GRCh38
rs748305262 Chr.21: 44287096-44287096 on Build GRCh38
rs368561485 Chr.21: 44287164-44287164 on Build GRCh38
rs1057516314 Chr.21: 44287127-44287128 on Build GRCh38
rs759776266 Chr.21: 44287198-44287198 on Build GRCh38
rs747601499 Chr.21: 44287122-44287122 on Build GRCh38
rs946115799 Chr.21: 44287191-44287191 on Build GRCh38
rs200377867 Chr.21: 44287127-44287127 on Build GRCh38
rs755258600 Chr.21: 44287094-44287094 on Build GRCh38
rs554372593 Chr.21: 44287123-44287123 on Build GRCh38
rs762735482 Chr.21: 44287057-44287057 on Build GRCh38
rs767362769 Chr.21: 44287151-44287151 on Build GRCh38
rs374549350 Chr.21: 44287157-44287157 on Build GRCh38
rs751460610 Chr.21: 44287081-44287081 on Build GRCh38
rs111925575 Chr.21: 44287219-44287219 on Build GRCh38
rs534476312 Chr.21: 44287199-44287199 on Build GRCh38
rs41276642 Chr.21: 44287098-44287098 on Build GRCh38
rs1030568996 Chr.21: 44287131-44287131 on Build GRCh38
rs1042807422 Chr.21: 44287089-44287089 on Build GRCh38
rs771341718 Chr.21: 44287116-44287116 on Build GRCh38
rs193922418 Chr.21: 44287133-44287133 on Build GRCh38
rs775210978 Chr.21: 44287125-44287125 on Build GRCh38
rs774680439 Chr.21: 44287100-44287100 on Build GRCh38
rs200023541 Chr.21: 44287178-44287178 on Build GRCh38
rs371165781 Chr.21: 44287154-44287154 on Build GRCh38
rs946198025 Chr.21: 44287046-44287046 on Build GRCh38
rs1022904624 Chr.21: 44287162-44287162 on Build GRCh38

View Full Product Details