Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 25469064-25469307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8A2
Entrez Gene ID
51761
Gene Name
ATPase phospholipid transporting 8A2
Gene Aliases
ATP, ATPIB, CAMRQ4, IB, ML-1
UniGene
Hs.444957
Gene Chromosome Location
Chr.13: 25372011-26025851 on Build GRCh38
Transcripts
NM_001313741.1 NM_016529.5 XM_005266419.1 XM_011535103.1 XM_011535104.2 XM_011535106.1 XM_011535107.2 XM_011535113.2 XM_017020625.1 XM_017020626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5639443 c.45C>T p.A15A Chr.13: 25469065-25469065 on Build GRCh38
5639442 c.165C>T p.A55A Chr.13: 25469065-25469065 on Build GRCh38
7127456 c.60C>T p.L20L Chr.13: 25469080-25469080 on Build GRCh38
7127455 c.180C>T p.L60L Chr.13: 25469080-25469080 on Build GRCh38
6074374 c.219C>A p.I73I Chr.13: 25469119-25469119 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs928970725 Chr.13: 25469203-25469203 on Build GRCh38
rs890961067 Chr.13: 25469218-25469218 on Build GRCh38
rs764713371 Chr.13: 25469124-25469126 on Build GRCh38
rs1001290877 Chr.13: 25469257-25469257 on Build GRCh38
rs12872195 Chr.13: 25469096-25469096 on Build GRCh38
rs200671371 Chr.13: 25469144-25469144 on Build GRCh38
rs368675559 Chr.13: 25469145-25469145 on Build GRCh38
rs1042333448 Chr.13: 25469235-25469235 on Build GRCh38
rs754364965 Chr.13: 25469105-25469105 on Build GRCh38
rs982197445 Chr.13: 25469089-25469089 on Build GRCh38
rs759915512 Chr.13: 25469155-25469155 on Build GRCh38
rs758765098 Chr.13: 25469168-25469168 on Build GRCh38
rs754335677 Chr.13: 25469153-25469153 on Build GRCh38
rs903728001 Chr.13: 25469254-25469254 on Build GRCh38
rs761517856 Chr.13: 25469084-25469084 on Build GRCh38
rs77815308 Chr.13: 25469267-25469267 on Build GRCh38
rs752999255 Chr.13: 25469162-25469162 on Build GRCh38
rs777981759 Chr.13: 25469115-25469115 on Build GRCh38
rs1051493657 Chr.13: 25469222-25469222 on Build GRCh38
rs12875883 Chr.13: 25469092-25469092 on Build GRCh38
rs370807035 Chr.13: 25469113-25469113 on Build GRCh38
rs770981523 Chr.13: 25469120-25469120 on Build GRCh38
rs183257147 Chr.13: 25469107-25469107 on Build GRCh38
rs747042311 Chr.13: 25469116-25469116 on Build GRCh38
rs533200780 Chr.13: 25469151-25469151 on Build GRCh38
rs765859369 Chr.13: 25469152-25469152 on Build GRCh38
rs749231912 Chr.13: 25469142-25469142 on Build GRCh38
rs113068536 Chr.13: 25469220-25469220 on Build GRCh38
rs1037547373 Chr.13: 25469146-25469146 on Build GRCh38
rs775484519 Chr.13: 25469140-25469140 on Build GRCh38
rs765753391 Chr.13: 25469159-25469159 on Build GRCh38
rs200891348 Chr.13: 25469148-25469148 on Build GRCh38
rs767302786 Chr.13: 25469086-25469086 on Build GRCh38
rs748540225 Chr.13: 25469108-25469108 on Build GRCh38
rs749896642 Chr.13: 25469090-25469090 on Build GRCh38
rs777027693 Chr.13: 25469162-25469163 on Build GRCh38
rs1050938596 Chr.13: 25469204-25469204 on Build GRCh38
rs546957142 Chr.13: 25469180-25469180 on Build GRCh38
rs370672539 Chr.13: 25469083-25469083 on Build GRCh38
rs766908597 Chr.13: 25469100-25469100 on Build GRCh38

View Full Product Details