Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44948474-44948745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOC2
Entrez Gene ID
344
Gene Name
apolipoprotein C2
Gene Aliases
APO-CII, APOC-II
UniGene
Hs.75615
Gene Chromosome Location
Chr.19: 44945982-44949565 on Build GRCh38
Transcripts
NM_000483.4 NR_037932.1

Gene Symbol
APOC4-APOC2
Entrez Gene ID
100533990
Gene Name
APOC4-APOC2 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.731599
Gene Chromosome Location
Chr.19: 44942238-44949565 on Build GRCh38
Transcripts
NM_000483.4 NR_037932.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5390086 c.4G>A p.G2S Chr.19: 44948482-44948482 on Build GRCh38
4891130 c.88G>A p.E30K Chr.19: 44948733-44948733 on Build GRCh38
5856131 c.5G>A p.G2D Chr.19: 44948483-44948483 on Build GRCh38
4953143 c.26T>C p.L9P Chr.19: 44948504-44948504 on Build GRCh38
6525101 c.76C>A p.P26T Chr.19: 44948721-44948721 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755352904 Chr.19: 44948501-44948502 on Build GRCh38
rs765634049 Chr.19: 44948681-44948682 on Build GRCh38
rs746636795 Chr.19: 44948549-44948549 on Build GRCh38
rs141530175 Chr.19: 44948497-44948497 on Build GRCh38
rs994371401 Chr.19: 44948713-44948713 on Build GRCh38
rs989170781 Chr.19: 44948610-44948610 on Build GRCh38
rs371548125 Chr.19: 44948653-44948653 on Build GRCh38
rs766438817 Chr.19: 44948658-44948658 on Build GRCh38
rs769051134 Chr.19: 44948551-44948551 on Build GRCh38
rs909824341 Chr.19: 44948500-44948500 on Build GRCh38
rs775028847 Chr.19: 44948547-44948547 on Build GRCh38
rs751858726 Chr.19: 44948591-44948591 on Build GRCh38
rs201709243 Chr.19: 44948703-44948703 on Build GRCh38
rs376235366 Chr.19: 44948660-44948660 on Build GRCh38
rs1039763443 Chr.19: 44948515-44948515 on Build GRCh38
rs757247241 Chr.19: 44948531-44948531 on Build GRCh38
rs750328283 Chr.19: 44948694-44948694 on Build GRCh38
rs778728968 Chr.19: 44948690-44948690 on Build GRCh38
rs114780592 Chr.19: 44948671-44948671 on Build GRCh38
rs370691343 Chr.19: 44948598-44948598 on Build GRCh38
rs969677284 Chr.19: 44948561-44948561 on Build GRCh38
rs1024038780 Chr.19: 44948695-44948695 on Build GRCh38
rs982676822 Chr.19: 44948700-44948700 on Build GRCh38
rs559469289 Chr.19: 44948542-44948542 on Build GRCh38
rs545692408 Chr.19: 44948538-44948538 on Build GRCh38
rs751752773 Chr.19: 44948502-44948502 on Build GRCh38
rs370703931 Chr.19: 44948550-44948550 on Build GRCh38
rs111628497 Chr.19: 44948534-44948534 on Build GRCh38
rs889880226 Chr.19: 44948675-44948675 on Build GRCh38
rs777678252 Chr.19: 44948514-44948514 on Build GRCh38
rs368074998 Chr.19: 44948592-44948592 on Build GRCh38
rs145690472 Chr.19: 44948527-44948527 on Build GRCh38
rs781495476 Chr.19: 44948620-44948620 on Build GRCh38
rs373756895 Chr.19: 44948678-44948678 on Build GRCh38
rs528176006 Chr.19: 44948643-44948643 on Build GRCh38
rs11553150 Chr.19: 44948519-44948519 on Build GRCh38
rs74500990 Chr.19: 44948697-44948697 on Build GRCh38
rs746715271 Chr.19: 44948705-44948705 on Build GRCh38
rs768288162 Chr.19: 44948708-44948708 on Build GRCh38
rs762369098 Chr.19: 44948573-44948573 on Build GRCh38
rs367610731 Chr.19: 44948540-44948540 on Build GRCh38
rs144145394 Chr.19: 44948718-44948718 on Build GRCh38
rs1130711 Chr.19: 44948715-44948715 on Build GRCh38
rs201090455 Chr.19: 44948670-44948670 on Build GRCh38
rs544199287 Chr.19: 44948501-44948501 on Build GRCh38
rs773707513 Chr.19: 44948654-44948654 on Build GRCh38
rs776961967 Chr.19: 44948562-44948562 on Build GRCh38
rs150887575 Chr.19: 44948512-44948512 on Build GRCh38
rs140169122 Chr.19: 44948518-44948518 on Build GRCh38
rs192252809 Chr.19: 44948563-44948563 on Build GRCh38

View Full Product Details