Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39101098-39101331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC3H
Entrez Gene ID
164668
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3H
Gene Aliases
A3H, ARP-10, ARP10
UniGene
Hs.440515
Gene Chromosome Location
Chr.22: 39097204-39104067 on Build GRCh38
Transcripts
NM_001166002.2 NM_001166003.2 NM_001166004.2 NM_181773.4 XM_011529990.2 XM_011529991.2 XM_011529992.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6602391 c.229A>G p.T77A Chr.22: 39101315-39101315 on Build GRCh38
6559403 c.182A>T p.N61I Chr.22: 39101268-39101268 on Build GRCh38
6602396 c.182A>G p.N61S Chr.22: 39101268-39101268 on Build GRCh38
1714402 c.184G>A p.E62K Chr.22: 39101270-39101270 on Build GRCh38
7150875 c.161A>G p.H54R Chr.22: 39101247-39101247 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757902029 Chr.22: 39101304-39101304 on Build GRCh38
rs904749051 Chr.22: 39101150-39101150 on Build GRCh38
rs924153597 Chr.22: 39101267-39101267 on Build GRCh38
rs141418864 Chr.22: 39101263-39101263 on Build GRCh38
rs745416513 Chr.22: 39101206-39101206 on Build GRCh38
rs897898864 Chr.22: 39101226-39101226 on Build GRCh38
rs776572204 Chr.22: 39101242-39101242 on Build GRCh38
rs768512741 Chr.22: 39101231-39101231 on Build GRCh38
rs991908437 Chr.22: 39101158-39101158 on Build GRCh38
rs567616108 Chr.22: 39101248-39101248 on Build GRCh38
rs138965536 Chr.22: 39101305-39101305 on Build GRCh38
rs778434817 Chr.22: 39101203-39101203 on Build GRCh38
rs950093697 Chr.22: 39101181-39101181 on Build GRCh38
rs530784331 Chr.22: 39101145-39101145 on Build GRCh38
rs199737025 Chr.22: 39101298-39101298 on Build GRCh38
rs370157569 Chr.22: 39101137-39101137 on Build GRCh38
rs771722034 Chr.22: 39101220-39101220 on Build GRCh38
rs954147247 Chr.22: 39101175-39101175 on Build GRCh38
rs767832239 Chr.22: 39101293-39101293 on Build GRCh38
rs761793621 Chr.22: 39101246-39101246 on Build GRCh38
rs1045840545 Chr.22: 39101204-39101204 on Build GRCh38
rs756103590 Chr.22: 39101223-39101223 on Build GRCh38
rs9611092 Chr.22: 39101176-39101176 on Build GRCh38
rs926211760 Chr.22: 39101153-39101153 on Build GRCh38
rs947813764 Chr.22: 39101294-39101294 on Build GRCh38
rs1022286301 Chr.22: 39101180-39101180 on Build GRCh38
rs745376430 Chr.22: 39101299-39101299 on Build GRCh38
rs145141547 Chr.22: 39101282-39101282 on Build GRCh38
rs770312650 Chr.22: 39101199-39101199 on Build GRCh38
rs567966449 Chr.22: 39101223-39101223 on Build GRCh38
rs200350520 Chr.22: 39101295-39101295 on Build GRCh38
rs13055594 Chr.22: 39101155-39101155 on Build GRCh38
rs866460154 Chr.22: 39101285-39101285 on Build GRCh38
rs968016551 Chr.22: 39101233-39101233 on Build GRCh38
rs570162336 Chr.22: 39101144-39101144 on Build GRCh38
rs866882348 Chr.22: 39101138-39101138 on Build GRCh38
rs766612664 Chr.22: 39101270-39101270 on Build GRCh38
rs765315072 Chr.22: 39101258-39101258 on Build GRCh38
rs748591691 Chr.22: 39101190-39101190 on Build GRCh38
rs763011975 Chr.22: 39101269-39101269 on Build GRCh38
rs751556615 Chr.22: 39101192-39101193 on Build GRCh38
rs929152741 Chr.22: 39101229-39101229 on Build GRCh38
rs760503179 Chr.22: 39101227-39101227 on Build GRCh38

View Full Product Details