Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153536389-153536614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2B3
Entrez Gene ID
492
Gene Name
ATPase plasma membrane Ca2+ transporting 3
Gene Aliases
CFAP39, CLA2, OPCA, PMCA3, PMCA3a, SCAX1
UniGene
Hs.533956
Gene Chromosome Location
Chr.X: 153517677-153582929 on Build GRCh38
Transcripts
NM_001001344.2 NM_021949.3 XM_005274689.3 XM_005274690.3 XM_005274691.3 XM_005274692.3 XM_011531174.2 XM_011531177.2 XM_017029553.1

SNP Detail

SNP ID Chromosome Location
rs912248707 Chr.X: 153536568-153536568 on Build GRCh38
rs1015450294 Chr.X: 153536445-153536445 on Build GRCh38
rs943758797 Chr.X: 153536577-153536577 on Build GRCh38
rs895866699 Chr.X: 153536428-153536428 on Build GRCh38
rs782391200 Chr.X: 153536477-153536477 on Build GRCh38
rs781927942 Chr.X: 153536507-153536507 on Build GRCh38
rs782444167 Chr.X: 153536438-153536438 on Build GRCh38
rs988566667 Chr.X: 153536581-153536581 on Build GRCh38
rs181673109 Chr.X: 153536463-153536463 on Build GRCh38
rs963559200 Chr.X: 153536448-153536448 on Build GRCh38
rs782018132 Chr.X: 153536494-153536494 on Build GRCh38
rs111567170 Chr.X: 153536549-153536549 on Build GRCh38
rs782804135 Chr.X: 153536557-153536557 on Build GRCh38
rs781933501 Chr.X: 153536534-153536534 on Build GRCh38

View Full Product Details