Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 988513-988738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP2A2
Entrez Gene ID
161
Gene Name
adaptor related protein complex 2 alpha 2 subunit
Gene Aliases
ADTAB, CLAPA2, HIP-9, HIP9, HYPJ
UniGene
Hs.19121
Gene Chromosome Location
Chr.11: 925809-1012245 on Build GRCh38
Transcripts
NM_001242837.1 NM_012305.3 XM_011519929.1 XM_011519930.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6070730 c.1159C>T p.R387W Chr.11: 988579-988579 on Build GRCh38
4037822 c.1193G>A p.R398H Chr.11: 988613-988613 on Build GRCh38
4462705 c.1254C>T p.S418S Chr.11: 988674-988674 on Build GRCh38
4523610 c.1219G>A p.E407K Chr.11: 988639-988639 on Build GRCh38
6849950 c.1253C>T p.S418F Chr.11: 988673-988673 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759104340 Chr.11: 988596-988596 on Build GRCh38
rs758908829 Chr.11: 988536-988536 on Build GRCh38
rs368388767 Chr.11: 988599-988599 on Build GRCh38
rs866811703 Chr.11: 988707-988707 on Build GRCh38
rs144441591 Chr.11: 988619-988619 on Build GRCh38
rs763234910 Chr.11: 988594-988595 on Build GRCh38
rs776381664 Chr.11: 988607-988607 on Build GRCh38
rs770771246 Chr.11: 988605-988605 on Build GRCh38
rs777411161 Chr.11: 988540-988540 on Build GRCh38
rs749816068 Chr.11: 988570-988570 on Build GRCh38
rs11538724 Chr.11: 988657-988657 on Build GRCh38
rs764823391 Chr.11: 988543-988543 on Build GRCh38
rs748221608 Chr.11: 988639-988639 on Build GRCh38
rs753815361 Chr.11: 988534-988534 on Build GRCh38
rs750808260 Chr.11: 988563-988563 on Build GRCh38
rs996958618 Chr.11: 988540-988541 on Build GRCh38
rs756614085 Chr.11: 988566-988566 on Build GRCh38
rs766218989 Chr.11: 988532-988532 on Build GRCh38
rs748276511 Chr.11: 988584-988584 on Build GRCh38
rs759551186 Chr.11: 988608-988608 on Build GRCh38
rs866531685 Chr.11: 988703-988703 on Build GRCh38
rs915678660 Chr.11: 988688-988688 on Build GRCh38
rs1001264133 Chr.11: 988549-988549 on Build GRCh38
rs751262548 Chr.11: 988620-988620 on Build GRCh38
rs762614728 Chr.11: 988615-988615 on Build GRCh38
rs765130178 Chr.11: 988609-988609 on Build GRCh38
rs1015727759 Chr.11: 988613-988613 on Build GRCh38
rs780490962 Chr.11: 988621-988621 on Build GRCh38
rs201537556 Chr.11: 988650-988650 on Build GRCh38
rs773847194 Chr.11: 988569-988569 on Build GRCh38
rs376073560 Chr.11: 988580-988580 on Build GRCh38
rs62640021 Chr.11: 988638-988638 on Build GRCh38
rs374078818 Chr.11: 988632-988632 on Build GRCh38
rs747250419 Chr.11: 988682-988682 on Build GRCh38
rs770589109 Chr.11: 988715-988715 on Build GRCh38
rs760392178 Chr.11: 988603-988603 on Build GRCh38
rs762196661 Chr.11: 988626-988626 on Build GRCh38
rs752237271 Chr.11: 988612-988612 on Build GRCh38
rs867485594 Chr.11: 988583-988583 on Build GRCh38
rs972990573 Chr.11: 988700-988700 on Build GRCh38
rs966764407 Chr.11: 988600-988600 on Build GRCh38
rs777620962 Chr.11: 988559-988559 on Build GRCh38
rs758095675 Chr.11: 988557-988557 on Build GRCh38
rs572932767 Chr.11: 988547-988547 on Build GRCh38
rs768577549 Chr.11: 988574-988574 on Build GRCh38
rs776458440 Chr.11: 988717-988717 on Build GRCh38

View Full Product Details