Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 27979666-27979933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF8L1
Entrez Gene ID
139425
Gene Name
DDB1 and CUL4 associated factor 8 like 1
Gene Aliases
WDR42B
UniGene
Hs.447365
Gene Chromosome Location
Chr.X: 27977993-27981449 on Build GRCh38
Transcripts
NM_001017930.1

SNP Detail

SNP ID Chromosome Location
rs755809602 Chr.X: 27979779-27979779 on Build GRCh38
rs1008975087 Chr.X: 27979778-27979778 on Build GRCh38
rs55924991 Chr.X: 27979749-27979749 on Build GRCh38
rs199848612 Chr.X: 27979699-27979699 on Build GRCh38
rs777469548 Chr.X: 27979795-27979795 on Build GRCh38
rs752458196 Chr.X: 27979764-27979764 on Build GRCh38
rs759385649 Chr.X: 27979698-27979698 on Build GRCh38
rs751954139 Chr.X: 27979751-27979751 on Build GRCh38
rs778425815 Chr.X: 27979868-27979868 on Build GRCh38
rs12388557 Chr.X: 27979690-27979690 on Build GRCh38
rs766931484 Chr.X: 27979734-27979734 on Build GRCh38
rs781689773 Chr.X: 27979760-27979760 on Build GRCh38
rs879063476 Chr.X: 27979786-27979786 on Build GRCh38
rs755400466 Chr.X: 27979757-27979757 on Build GRCh38
rs138100395 Chr.X: 27979716-27979716 on Build GRCh38
rs771911063 Chr.X: 27979888-27979888 on Build GRCh38
rs770284640 Chr.X: 27979798-27979798 on Build GRCh38
rs762007041 Chr.X: 27979702-27979702 on Build GRCh38
rs368290304 Chr.X: 27979710-27979710 on Build GRCh38
rs745341683 Chr.X: 27979839-27979839 on Build GRCh38
rs765350190 Chr.X: 27979704-27979704 on Build GRCh38
rs774103564 Chr.X: 27979689-27979689 on Build GRCh38
rs745785986 Chr.X: 27979876-27979876 on Build GRCh38

View Full Product Details