Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44689499-44689744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf228
Entrez Gene ID
339541
Gene Name
chromosome 1 open reading frame 228
Gene Aliases
NCRNA00082, p40
UniGene
Hs.173679
Gene Chromosome Location
Chr.1: 44674696-44725591 on Build GRCh38
Transcripts
NM_001145636.1 XM_006710603.2 XM_006710604.2 XM_011541340.1 XM_011541341.1 XM_011541343.1 XM_011541345.1 XM_011541349.1 XM_011541354.1 XM_011541355.1 XM_017001140.1 XM_017001141.1 XM_017001142.1 XM_017001143.1 XM_017001144.1 XM_017001145.1 XM_017001146.1 XM_017001147.1 XM_017001148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5381089 c.32G>C p.S11T Chr.1: 44689729-44689729 on Build GRCh38
5478699 c.6T>C p.T2T Chr.1: 44689703-44689703 on Build GRCh38
5823793 c.46T>G p.F16V Chr.1: 44689743-44689743 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747080049 Chr.1: 44689553-44689553 on Build GRCh38
rs906651145 Chr.1: 44689671-44689671 on Build GRCh38
rs3795710 Chr.1: 44689537-44689537 on Build GRCh38
rs976510679 Chr.1: 44689527-44689527 on Build GRCh38
rs945342550 Chr.1: 44689623-44689623 on Build GRCh38
rs16832188 Chr.1: 44689554-44689554 on Build GRCh38
rs148467508 Chr.1: 44689549-44689549 on Build GRCh38
rs1046484279 Chr.1: 44689644-44689644 on Build GRCh38
rs549594321 Chr.1: 44689530-44689530 on Build GRCh38
rs747547395 Chr.1: 44689556-44689556 on Build GRCh38
rs775050209 Chr.1: 44689705-44689705 on Build GRCh38
rs892477138 Chr.1: 44689597-44689597 on Build GRCh38
rs3795711 Chr.1: 44689538-44689538 on Build GRCh38
rs771559418 Chr.1: 44689696-44689696 on Build GRCh38
rs1033848163 Chr.1: 44689707-44689707 on Build GRCh38
rs942754257 Chr.1: 44689524-44689524 on Build GRCh38
rs746131170 Chr.1: 44689657-44689657 on Build GRCh38
rs1034983995 Chr.1: 44689567-44689567 on Build GRCh38
rs535968973 Chr.1: 44689522-44689522 on Build GRCh38

View Full Product Details