Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169106855-169107102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1B1
Entrez Gene ID
481
Gene Name
ATPase Na+/K+ transporting subunit beta 1
Gene Aliases
ATP1B
UniGene
Hs.291196
Gene Chromosome Location
Chr.1: 169106709-169132722 on Build GRCh38
Transcripts
NM_001677.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5477785 c.51G>A p.W17* Chr.1: 169106880-169106880 on Build GRCh38
463360 c.78C>T p.G26G Chr.1: 169106907-169106907 on Build GRCh38
1744859 c.59_61delAGA p.K22delK Chr.1: 169106888-169106890 on Build GRCh38
5952163 c.97+2_97+3insT p.? Chr.1: 169106928-169106929 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs999822367 Chr.1: 169106944-169106944 on Build GRCh38
rs775861246 Chr.1: 169106890-169106890 on Build GRCh38
rs746063893 Chr.1: 169107076-169107076 on Build GRCh38
rs534114378 Chr.1: 169106955-169106955 on Build GRCh38
rs766416126 Chr.1: 169106940-169106940 on Build GRCh38
rs544745237 Chr.1: 169107006-169107006 on Build GRCh38
rs755922216 Chr.1: 169106971-169106971 on Build GRCh38
rs147872554 Chr.1: 169106898-169106898 on Build GRCh38
rs776974246 Chr.1: 169106926-169106926 on Build GRCh38
rs770432036 Chr.1: 169106888-169106888 on Build GRCh38
rs770833468 Chr.1: 169106935-169106935 on Build GRCh38
rs200590991 Chr.1: 169106972-169106972 on Build GRCh38
rs773375182 Chr.1: 169106939-169106939 on Build GRCh38
rs1200128 Chr.1: 169106974-169106974 on Build GRCh38
rs751470349 Chr.1: 169106950-169106950 on Build GRCh38
rs955774598 Chr.1: 169106963-169106963 on Build GRCh38
rs768959839 Chr.1: 169106907-169106907 on Build GRCh38
rs975030649 Chr.1: 169107000-169107000 on Build GRCh38
rs909912701 Chr.1: 169107034-169107034 on Build GRCh38
rs778473563 Chr.1: 169106978-169106978 on Build GRCh38
rs376416247 Chr.1: 169106937-169106937 on Build GRCh38
rs1048419055 Chr.1: 169107025-169107025 on Build GRCh38
rs574108307 Chr.1: 169107040-169107040 on Build GRCh38
rs930585645 Chr.1: 169107007-169107007 on Build GRCh38
rs1037138263 Chr.1: 169107055-169107055 on Build GRCh38
rs551536131 Chr.1: 169106883-169106883 on Build GRCh38
rs879136253 Chr.1: 169106913-169106913 on Build GRCh38
rs956769261 Chr.1: 169107012-169107012 on Build GRCh38
rs1023231060 Chr.1: 169106934-169106934 on Build GRCh38
rs368987392 Chr.1: 169106956-169106956 on Build GRCh38
rs989461123 Chr.1: 169107016-169107016 on Build GRCh38
rs555887014 Chr.1: 169106986-169106986 on Build GRCh38
rs752511552 Chr.1: 169106967-169106967 on Build GRCh38
rs965917093 Chr.1: 169106992-169106992 on Build GRCh38
rs554366516 Chr.1: 169107026-169107026 on Build GRCh38
rs879113954 Chr.1: 169106927-169106927 on Build GRCh38
rs577653520 Chr.1: 169106988-169106988 on Build GRCh38
rs61204485 Chr.1: 169106962-169106962 on Build GRCh38
rs543354960 Chr.1: 169107084-169107084 on Build GRCh38

View Full Product Details