Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128167020-128167251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIZ1
Entrez Gene ID
25792
Gene Name
CDKN1A interacting zinc finger protein 1
Gene Aliases
LSFR1, NP94, ZNF356
UniGene
Hs.212395
Gene Chromosome Location
Chr.9: 128166065-128204383 on Build GRCh38
Transcripts
NM_001131015.1 NM_001131016.1 NM_001131017.1 NM_001131018.1 NM_001257975.1 NM_001257976.1 NM_012127.2 XM_005251888.3 XM_005251891.3 XM_005251892.3 XM_005251893.4 XM_006717039.3 XM_011518481.1 XM_011518482.2 XM_011518483.1 XM_017014594.1 XM_017014595.1 XM_017014596.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1105491 c.2337G>A p.S779S Chr.9: 128167123-128167123 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761070181 Chr.9: 128167214-128167214 on Build GRCh38
rs200395357 Chr.9: 128167124-128167124 on Build GRCh38
rs533825245 Chr.9: 128167225-128167225 on Build GRCh38
rs200887788 Chr.9: 128167047-128167047 on Build GRCh38
rs758057153 Chr.9: 128167189-128167189 on Build GRCh38
rs781644311 Chr.9: 128167139-128167150 on Build GRCh38
rs931173760 Chr.9: 128167197-128167197 on Build GRCh38
rs889552362 Chr.9: 128167215-128167215 on Build GRCh38
rs1010665562 Chr.9: 128167224-128167224 on Build GRCh38
rs201027257 Chr.9: 128167146-128167146 on Build GRCh38
rs780540900 Chr.9: 128167160-128167160 on Build GRCh38
rs375396341 Chr.9: 128167175-128167175 on Build GRCh38
rs142689254 Chr.9: 128167106-128167106 on Build GRCh38
rs893600885 Chr.9: 128167222-128167222 on Build GRCh38
rs372111295 Chr.9: 128167174-128167174 on Build GRCh38
rs773718838 Chr.9: 128167194-128167194 on Build GRCh38
rs947936704 Chr.9: 128167171-128167171 on Build GRCh38
rs767974380 Chr.9: 128167092-128167092 on Build GRCh38
rs999081310 Chr.9: 128167091-128167091 on Build GRCh38
rs769738421 Chr.9: 128167164-128167164 on Build GRCh38
rs1049529845 Chr.9: 128167208-128167208 on Build GRCh38
rs779920513 Chr.9: 128167173-128167173 on Build GRCh38
rs571270532 Chr.9: 128167190-128167190 on Build GRCh38
rs199622355 Chr.9: 128167127-128167127 on Build GRCh38
rs756753256 Chr.9: 128167123-128167123 on Build GRCh38
rs764248899 Chr.9: 128167080-128167080 on Build GRCh38
rs957561325 Chr.9: 128167120-128167120 on Build GRCh38
rs563232675 Chr.9: 128167144-128167144 on Build GRCh38
rs746724700 Chr.9: 128167135-128167135 on Build GRCh38
rs375305609 Chr.9: 128167227-128167227 on Build GRCh38
rs762965441 Chr.9: 128167064-128167064 on Build GRCh38
rs183958510 Chr.9: 128167087-128167087 on Build GRCh38
rs935179221 Chr.9: 128167201-128167201 on Build GRCh38
rs540341207 Chr.9: 128167058-128167058 on Build GRCh38
rs1052174395 Chr.9: 128167220-128167220 on Build GRCh38
rs752276279 Chr.9: 128167126-128167126 on Build GRCh38
rs745859418 Chr.9: 128167056-128167056 on Build GRCh38
rs774596799 Chr.9: 128167082-128167082 on Build GRCh38
rs369422393 Chr.9: 128167182-128167182 on Build GRCh38

View Full Product Details